Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01260127_m1
          See other 1700003I22RIK GE Assays ›
          Gene Symbol:
          1700003I22Rik
          Gene Name:
          RIKEN cDNA 1700003I22 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 56018978 - 56020203 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01260127_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700003I22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK005643.1 - 1 - 2 81 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01075850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20244990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20244997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00934968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01078403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00672423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00672422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01007035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20244992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00815692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00672409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00609860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00609861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00815695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00065476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00934967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00672414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20244998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00386496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00806919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00020768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00386490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00575856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20245058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00672421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20244994 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 169169_PM_at - -
          Murine Genome U74v2 Set 169169_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763359 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102030672 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline