Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01260550_g1
          See other IFI35 GE Assays ›
          Gene Symbol:
          Ifi35
          Gene Name:
          interferon-induced protein 35
          Gene Aliases:
          2010008K16Rik, AW986054, IFP35, ifi-35
          Chromosome Location:
          Chr.11: 101448412 - 101458701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01260550_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi35
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027320.4 NP_081596.1 2 - 3 311 118
          GenBank mRNA AK008161.1 - 2 - 3 243 118
          AK131884.1 - 2 - 3 312 118
          BC008158.1 - 2 - 3 215 3155505 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001657.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifi35

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 BC008158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 AK008161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 AK131884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 NM_027320.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 AK154178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 BC008158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 AK008161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 AK131884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 NM_027320.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P414889 AK154178.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 196324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159507 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105335 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 BC008158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 AK008161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 AK131884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 NM_027320.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 AK154178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 BC008158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 AK008161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 AK131884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 NM_027320.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P414889 AK154178.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073935 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02411723 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07466689 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105335 - -
          Whole Mouse Genome G4122A A_51_P414889 BC008158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 AK008161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 AK131884.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 NM_027320.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 AK154178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 BC008158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 AK008161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 AK131884.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 NM_027320.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P414889 AK154178.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135015 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105335 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09735464 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09735466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09735462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28074035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09208665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09338886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09027896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09523092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09735466 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381415 - -
          Affymetrix HT MG-430 PM Array Plate 1424617_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1445897_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1459151_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381408 - -
          Mouse Genome 430 2.0 Array 1424617_at - -
          Mouse Genome 430 2.0 Array 1445897_s_at - -
          Mouse Genome 430 2.0 Array 1445897_PM_s_at - -
          Mouse Genome 430 2.0 Array 1459151_x_at - -
          Mouse Genome 430 2.0 Array 1424617_PM_at - -
          Mouse Genome 430 2.0 Array 1459151_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70110_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70110_at - -
          Affymetrix Mouse Clariom S Array PSR1100015532.mm.1 NM_027320.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015532.mm.1 NM_027320.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015532.mm.1 TC1100001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015540.mm.1 NM_027320.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015540.mm.1 NM_027320.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015540.mm.1 TC1100001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015534.mm.1 NM_027320.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015534.mm.1 NM_027320.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015534.mm.1 TC1100001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015535.mm.1 NM_027320.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015535.mm.1 NM_027320.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015535.mm.1 TC1100001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015537.mm.1 NM_027320.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015537.mm.1 NM_027320.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015537.mm.1 TC1100001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015541.mm.1 NM_027320.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015541.mm.1 NM_027320.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015541.mm.1 TC1100001657.mm.1 MTA TC
          Mouse Expression Set 430 1424617_PM_at - -
          Mouse Expression Set 430 1424617_at - -
          Murine Genome U74v2 Set 100013_PM_at - -
          Murine Genome U74v2 Set 100013_at - -
          Mouse Genome 430A 2.0 Array 1424617_PM_at - -
          Mouse Genome 430A 2.0 Array 1424617_at - -
          Affymetrix HT Mouse Genome 430A Array 1424617_at - -
          Affymetrix HT Mouse Genome 430A Array 1424617_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459151_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1445897_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1459151_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1445897_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 546373 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230739 - -
          Illumina mouse-6 v1.0 expression beadchip 4230739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625290 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625290 - -
          mouse-6 v1.1 (Illumina) scl40880.4.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40880.4.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625290 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625290 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40880.4.1_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline