Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01262543_m1
          See other POLI GE Assays ›
          Gene Symbol:
          Poli
          Gene Name:
          polymerase (DNA directed), iota
          Gene Aliases:
          Rad30b
          Chromosome Location:
          Chr.18: 70508680 - 70530334 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01262543_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Poli
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136090.2 NP_001129562.1 2 - 3 700 75
          NM_001289515.1 NP_001276444.1 2 - 3 700 75
          NM_001289516.1 NP_001276445.1 2 - 3 700 75
          NM_011972.2 NP_036102.2 2 - 3 263 75
          GenBank mRNA AF151691.1 - 2 - 3 263 75
          AK145244.1 - 2 - 3 208 75
          AK154537.1 - 2 - 3 649 75
          AK163784.1 - 2 - 3 221 75
          AY515316.1 - 2 - 3 169 75
          AY515317.1 - 2 - 3 169 75
          BC057575.1 - 2 - 3 660 75
          BC082278.1 - 2 - 3 517 6328261 75
          HQ698885.1 - 2 - 3 396 75
          HQ698886.1 - 2 - 3 396 75
          HQ698887.1 - 2 - 3 396 75
          HQ698888.1 - 2 - 3 396 75
          HQ698889.1 - 2 - 3 396 75
          HQ698890.1 - 2 - 3 396 75
          HQ698891.1 - 2 - 3 396 75
          HQ698892.1 - 2 - 3 396 75
          HQ698893.1 - 2 - 3 396 75

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK154537.1, BC082278.1, NM_001136090.2, NM_001289515.1, NM_001289516.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Poli

          Panther Classification:

          Molecular Function -

          DNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA repair
          cellular response to DNA damage stimulus
          translesion synthesis
          cellular response to UV-C
          DNA biosynthetic process
          DNA binding
          damaged DNA binding
          DNA-directed DNA polymerase activity
          transferase activity
          nucleotidyltransferase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800011296.mm.1 NM_001136090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011296.mm.1 NM_001289515.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011296.mm.1 NM_001289516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011296.mm.1 TC1800001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011293.mm.1 NM_001136090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011293.mm.1 NM_001289515.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011293.mm.1 NM_001289516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011293.mm.1 NM_011972.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011293.mm.1 NM_011972.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011293.mm.1 TC1800001541.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011297.mm.1 NM_001136090.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011297.mm.1 NM_001289515.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011297.mm.1 NM_001289516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011297.mm.1 TC1800001541.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline