Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264676_m1
          See other PLAC8L1 GE Assays ›
          Gene Symbol:
          Plac8l1
          Gene Name:
          PLAC8-like 1
          Gene Aliases:
          1700009N18Rik, 1700015M15Rik
          Chromosome Location:
          Chr.18: 42178675 - 42196709 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264676_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plac8l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027072.1 NP_081348.1 3 - 4 477 64
          GenBank mRNA AK006002.1 - 3 - 4 455 64
          BC110558.1 - 3 - 4 439 40049443 64
          BC110559.1 - 3 - 4 439 40049451 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001253.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 BC110559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 NM_027072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 AK006002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 BC110558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 BC110559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 NM_027072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 AK006002.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P335993 BC110558.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 364665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 643036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 210300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24035 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23950 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178583 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154085 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 BC110559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 NM_027072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 AK006002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 BC110558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 BC110559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 NM_027072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 AK006002.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P335993 BC110558.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08978875 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154085 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178583 - -
          Whole Mouse Genome G4122A A_51_P335993 BC110559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 NM_027072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 AK006002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 BC110558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 BC110559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 NM_027072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 AK006002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335993 BC110558.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107947 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178583 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13768028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14031927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13812439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13768041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13768015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13967468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14064571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13768018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13639645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31654122 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458605 - -
          Affymetrix HT MG-430 PM Array Plate 1447794_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1429892_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458601 - -
          Mouse Genome 430 2.0 Array 1429892_at - -
          Mouse Genome 430 2.0 Array 1429892_PM_at - -
          Mouse Genome 430 2.0 Array 1447794_PM_x_at - -
          Mouse Genome 430 2.0 Array 1447794_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69401_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69401_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800009285.mm.1 NM_027072.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009285.mm.1 NM_027072.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009285.mm.1 TC1800001253.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009284.mm.1 NM_027072.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009284.mm.1 NM_027072.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009284.mm.1 TC1800001253.mm.1 MTA TC
          Mouse Expression Set 430 1447794_PM_x_at - -
          Mouse Expression Set 430 1429892_PM_at - -
          Mouse Expression Set 430 1447794_x_at - -
          Mouse Expression Set 430 1429892_at - -
          Murine Genome U74v2 Set 167411_PM_i_at - -
          Murine Genome U74v2 Set 167411_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1429892_at - -
          Affymetrix HT Mouse Genome 430B Array 1447794_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1429892_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447794_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884608 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215532 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline