Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264871_m1
          See other 8030443G20RIK GE Assays ›
          Gene Symbol:
          8030443G20Rik
          Gene Name:
          RIKEN cDNA 8030443G20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 108942843 - 108972077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264871_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 8030443G20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040664.1 3 - 4 394 95
          GenBank mRNA AK020199.1 - 3 - 4 395 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003258.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520315 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173974 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148877 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148877 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00957291 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00957289 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140789 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134366 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23198524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23198528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23198520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03806869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03568789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23198525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23198527 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400025917.mm.1 NR_040664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025917.mm.1 NR_040664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025917.mm.1 TC0400003258.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400025916.mm.1 NR_040664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025916.mm.1 NR_040664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025916.mm.1 TC0400003258.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400025918.mm.1 NR_040664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025918.mm.1 NR_040664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025918.mm.1 TC0400003258.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400025915.mm.1 NR_040664.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025915.mm.1 NR_040664.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400025915.mm.1 TC0400003258.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507014 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106620095 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline