Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01266279_g1
          See other MMP23 GE Assays ›
          Gene Symbol:
          Mmp23
          Gene Name:
          matrix metallopeptidase 23
          Gene Aliases:
          CA-MMP
          Chromosome Location:
          Chr.4: 155650655 - 155654481 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01266279_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmp23
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011985.2 NP_036115.1 3 - 4 638 62
          GenBank mRNA AF085742.1 - 3 - 4 638 62
          AK146510.1 - 3 - 4 445 62
          BC107357.1 - 3 - 4 545 40055939 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004146.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmp23

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          metalloendopeptidase activity
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439842 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439844 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439845 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 BC107357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 BC107358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 NM_011985.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 AF085742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 AK146510.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 BC107357.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 BC107358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 NM_011985.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 AF085742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P502132 AK146510.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866962 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137854 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 BC107357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 BC107358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 NM_011985.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 AF085742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 AK146510.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 BC107357.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 BC107358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 NM_011985.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 AF085742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P502132 AK146510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 BC107357.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 BC107358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 NM_011985.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 AF085742.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 AK146510.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 BC107357.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 BC107358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 NM_011985.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 AF085742.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502132 AK146510.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23439840 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137854 - -
          Whole Mouse Genome G4122A A_51_P502132 BC107357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 BC107358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 NM_011985.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 AF085742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 AK146510.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 BC107357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 BC107358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 NM_011985.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 AF085742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502132 AK146510.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140150 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137854 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056497 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056504 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04056557 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439842 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439844 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23439845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23439851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03254382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04056497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04056537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04056554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04056525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23439819 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519144 - -
          Affymetrix HT MG-430 PM Array Plate 1417282_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417281_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519140 - -
          Mouse Genome 430 2.0 Array 1417281_PM_a_at - -
          Mouse Genome 430 2.0 Array 1417282_PM_at - -
          Mouse Genome 430 2.0 Array 1417282_at - -
          Mouse Genome 430 2.0 Array 1417281_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26561_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26561_at - -
          Affymetrix Mouse Clariom S Array PSR0400034751.mm.1 TC0400004146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400034751.mm.1 NM_011985.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400034749.mm.1 TC0400004146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400034749.mm.1 NM_011985.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400034750.mm.1 TC0400004146.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400034750.mm.1 NM_011985.3 Refseq
          Mouse Expression Set 430 1417282_PM_at - -
          Mouse Expression Set 430 1417281_PM_a_at - -
          Mouse Expression Set 430 1417282_at - -
          Mouse Expression Set 430 1417281_a_at - -
          Mouse Genome 430A 2.0 Array 1417281_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417281_a_at - -
          Mouse Genome 430A 2.0 Array 1417282_at - -
          Mouse Genome 430A 2.0 Array 1417282_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417282_at - -
          Affymetrix HT Mouse Genome 430A Array 1417281_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417281_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417282_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 623672 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3940278 - -
          Illumina mouse-6 v1.0 expression beadchip 3940278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237364 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237364 - -
          mouse-6 v1.1 (Illumina) scl23434.8.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23434.8.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237364 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline