Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01267656_s1
          See other KLHL9 GE Assays ›
          Gene Symbol:
          Klhl9
          Gene Name:
          kelch-like 9
          Gene Aliases:
          8030469P05, C530050O22Rik, ENSMUSG00000070923, mKIAA1354
          Chromosome Location:
          Chr.4: 88718292 - 88722508 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01267656_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klhl9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172871.2 NP_766459.2 1 3843 98
          GenBank mRNA AK129338.1 - 1 3754 98

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klhl9

          Gene Ontology Categories:

          Function(s) Process(es)

          cytokinesis
          cell cycle
          mitotic nuclear division
          protein ubiquitination
          cell division
          ubiquitin-protein transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091885 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091881 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091879 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091883 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154714 AK129338.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2154714 NM_172871.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391384 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106664 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154714 AK129338.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2154714 NM_172871.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091891 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23091864 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106664 - -
          Whole Mouse Genome G4122A A_52_P1083725 AK144774.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P567839 AK049803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P567839 AK129338.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P567839 NM_172871.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83615 AK049803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83615 AK129338.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83615 BC094556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83615 AK033231.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P83615 NM_172871.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113824 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03917226 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03296779 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03296760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03917223 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03296774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03917227 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03917225 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03296759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03917231 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091881 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091879 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23091885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23091811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03364799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03388987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03430420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03388988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03917222 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03364800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23091916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03296759 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514320 - -
          Affymetrix HT MG-430 PM Array Plate 1433442_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514319 - -
          Affymetrix Mouse Gene 1.1 ST Array 10505839 - -
          Mouse Genome 430 2.0 Array 1433442_at - -
          Mouse Genome 430 2.0 Array 1433442_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242521_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242521_at - -
          Affymetrix HT Mouse Genome 430A Array 1433442_at - -
          Affymetrix HT Mouse Genome 430A Array 1433442_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 502640 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690348 - -
          Illumina mouse-6 v1.0 expression beadchip 1660068 - -
          Illumina mouse-6 v1.0 expression beadchip 5690348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246060 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2592779 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2592779 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246060 - -
          mouse-6 v1.1 (Illumina) scl00242521.1_200-S - -
          mouse-6 v1.1 (Illumina) scl24128.1.6_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660068 - -
          Illumina Sentrix mouse-6.v1 bead array 5690348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00242521.1_200-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24128.1.6_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246060 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2592779 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246060 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2592779 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00242521.1_200-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24128.1.6_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0