Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01269242_m1
          See other OARD1 GE Assays ›
          Gene Symbol:
          Oard1
          Gene Name:
          O-acyl-ADP-ribose deacylase 1
          Gene Aliases:
          AI314976, AW558560
          Chromosome Location:
          Chr.17: 48409985 - 48417270 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01269242_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oard1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001289490.1 NP_001276419.1 3 - 4 588 101
          NM_001289491.1 NP_001276420.1 3 - 4 313 101
          NM_207219.4 NP_997102.2 3 - 4 453 101
          GenBank mRNA AK148136.1 - 3 - 4 575 101
          AK153512.1 - 3 - 4 376 101
          BC022574.1 - 2 - 3 232 3989373 101
          BC157987.1 - 3 - 4 338 9007303 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000939.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oard1

          Gene Ontology Categories:

          Function(s) Process(es)

          purine nucleoside metabolic process
          purine nucleoside binding
          histone deacetylase activity
          protein-N-terminal asparagine amidohydrolase activity
          UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
          hydrolase activity
          iprodione amidohydrolase activity
          (3,5-dichlorophenylurea)acetate amidohydrolase activity
          deacetylase activity
          4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
          didemethylisoproturon amidohydrolase activity
          N-isopropylacetanilide amidohydrolase activity
          N-cyclohexylformamide amidohydrolase activity
          isonicotinic acid hydrazide hydrolase activity
          cis-aconitamide amidase activity
          gamma-N-formylaminovinylacetate hydrolase activity
          N2-acetyl-L-lysine deacetylase activity
          O-succinylbenzoate synthase activity
          indoleacetamide hydrolase activity
          N-acetylcitrulline deacetylase activity
          N-acetylgalactosamine-6-phosphate deacetylase activity
          diacetylchitobiose deacetylase activity
          chitooligosaccharide deacetylase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31217297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31217298 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408561 AK153512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408561 BC022574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408561 NM_207219.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408561 AK153512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408561 BC022574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408561 NM_207219.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 83135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485453 XM_357199.4 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485453 XM_909909.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110171 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155915 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164661 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153444 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408561 AK153512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408561 BC022574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408561 NM_207219.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408561 AK153512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408561 BC022574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408561 NM_207219.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31217300 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155915 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153444 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164661 - -
          Whole Mouse Genome G4122A A_52_P505584 AK148136.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P505584 AK153512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P505584 BC022574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P505584 BC157987.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P505584 NM_207219.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408561 AK153512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408561 BC022574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408561 NM_207219.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408561 AK153512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408561 BC022574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408561 NM_207219.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115456 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153444 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164661 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13472843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13472856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13472847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31217297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31217298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31217379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13198418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13238601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13472847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13238613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13238587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13238595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13238607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31217304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445801 - -
          Affymetrix HT MG-430 PM Array Plate 1449614_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445796 - -
          Mouse Genome 430 2.0 Array 1449614_s_at - -
          Mouse Genome 430 2.0 Array 1449614_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106821_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106821_at - -
          Affymetrix Mouse Clariom S Array PSR1700009081.mm.1 NM_001289490.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009081.mm.1 NM_001289491.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009081.mm.1 NM_207219.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009081.mm.1 TC1700000939.mm.1 MTA TC
          Mouse Expression Set 430 1449614_s_at - -
          Mouse Expression Set 430 1449614_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449614_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449614_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449614_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449614_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 338040 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3830280 - -
          Illumina mouse-6 v1.0 expression beadchip 3830280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830280 - -
          mouse-6 v1.1 (Illumina) scl50630.6.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50630.6.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2942000 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2638036 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2942000 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline