Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01291559_m1
          See other GM14635 GE Assays ›
          Gene Symbol:
          Gm14635
          Gene Name:
          predicted gene 14635
          Gene Aliases:
          OTTMUSG00000017536
          Chromosome Location:
          Chr.X: 12339779 - 12354996 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01291559_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14635
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045321.1 3 - 4 332 137
          GenBank mRNA AK075658.1 - 3 - 4 333 137

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P133112 AK075658.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 282373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213809 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P133112 AK075658.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03641342 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133112 AK075658.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P14523761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14790995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14612527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15143399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14784415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14569648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14973052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14523763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14523759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14712593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14583978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14818443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14569644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32261669 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline