Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01294625_s1
          See other OLFR1414 GE Assays ›
          Gene Symbol:
          Olfr1414
          Gene Name:
          olfactory receptor 1414
          Gene Aliases:
          MOR103-2
          Chromosome Location:
          Chr.1: 92511048 - 92518515 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01294625_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1414
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_147039.2 NP_667250.1 2 420 79
          GenBank mRNA BC120830.1 - 1 247 40129753 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1414

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P330452 NM_147039.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P330452 BC120830.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P330452 NM_147039.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P330452 BC120830.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106420 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P330452 NM_147039.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P330452 BC120830.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P330452 NM_147039.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P330452 BC120830.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P330452 NM_147039.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P330452 BC120830.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P330452 NM_147039.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P330452 BC120830.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20449530 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106420 - -
          Whole Mouse Genome G4122A A_51_P330452 NM_147039.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P330452 BC120830.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P330452 NM_147039.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P330452 BC120830.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124488 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112134 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106420 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00838414 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00838427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00838406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00838398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20449540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00526451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00165686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00838417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00343875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00838428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00159630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00316878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01131339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00316886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00165691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00838429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00207760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00207761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00749851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00804645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01030034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20449489 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356676 - -
          Affymetrix Mouse Gene 1.1 ST Array 10356675 - -
          Affymetrix Mouse Gene 1.1 ST Array 10356673 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 695265 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6100168 - -
          Illumina mouse-6 v1.0 expression beadchip 6100168 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100168 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222406 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222406 - -
          mouse-6 v1.1 (Illumina) scl16470.1.1_255-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100168 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16470.1.1_255-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100168 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222406 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222406 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0