Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01297279_m1
          See other SPAG9 GE Assays ›
          Gene Symbol:
          Spag9
          Gene Name:
          sperm associated antigen 9
          Gene Aliases:
          3110018C07Rik, 4733401I23Rik, 4831406C20Rik, AW552012, JLP, JSAP2, JSAP2a, Jip4, Mapk8ip4, syd1
          Chromosome Location:
          Chr.11: 93996058 - 94126087 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01297279_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spag9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025428.1 NP_001020599.1 25 - 26 3401 64
          NM_001025429.1 NP_001020600.1 25 - 26 3398 64
          NM_001025430.1 NP_001020601.1 24 - 25 3386 64
          NM_001199203.1 NP_001186132.1 28 - 29 3877 64
          NM_001199204.1 NP_001186133.1 28 - 29 3874 64
          NM_001199205.1 NP_001186134.1 25 - 26 3425 64
          NM_027569.2 NP_081845.2 27 - 28 3835 64
          GenBank mRNA AB047782.1 - 28 - 29 3797 64
          AF327451.1 - 27 - 28 3817 64
          AK049161.1 - 19 - 20 2437 64
          AK147431.1 - 25 - 26 3482 64
          AK147537.1 - 25 - 26 3501 64
          AK172961.1 - 25 - 26 3380 64
          AY823270.1 - 27 - 28 3379 64
          BC052058.1 - 25 - 26 3336 64
          BC060100.1 - 24 - 25 3320 6417257 64
          BC060506.1 - 13 - 14 1758 64
          BC094670.1 - 24 - 25 3314 30618738 64
          FJ042496.1 - 28 - 29 3751 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001425.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spag9

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          activation of JUN kinase activity
          positive regulation of signal transduction
          vesicle-mediated transport
          positive regulation of cell migration
          retrograde transport, endosome to Golgi
          positive regulation of MAPK cascade
          positive regulation of neuron differentiation
          striated muscle cell differentiation
          protein homooligomerization
          negative regulation of protein homodimerization activity
          MAP-kinase scaffold activity
          protein binding
          JUN kinase binding
          kinesin binding
          receptor signaling complex scaffold activity
          mitogen-activated protein kinase p38 binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001025428.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001025428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001025429.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001025429.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001025430.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001025430.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001199203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001199203.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001199204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001199204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001199205.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_001199205.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_027569.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 NM_027569.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100013300.mm.1 TC1100001425.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline