Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pnldc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034866.1 | NP_001030038.1 | 14 - 15 | 1200 | 66 | |
GenBank mRNA | BC158033.1 | - | 14 - 15 | 1200 | 9007336 | 66 |
BC172002.1 | - | 14 - 15 | 1203 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |