Search
Search

Cenpj| Gene Symbol: Cenpj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014996.2 | NP_001014996.2 | 8 - 9 | 2998 | 77 | |
| GenBank mRNA | AK077045.1 | - | 8 - 9 | 2981 | 77 | |
| BC092399.1 | - | 8 - 9 | 2984 | 30549055 | 77 | |
Set Membership: |
|