Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01349164_g1
          See other ARRDC2 GE Assays ›
          Gene Symbol:
          Arrdc2
          Gene Name:
          arrestin domain containing 2
          Gene Aliases:
          4632416I05Rik, Ilad1
          Chromosome Location:
          Chr.8: 70835129 - 70839720 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01349164_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arrdc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027560.1 NP_081836.1 7 - 8 1492 51
          GenBank mRNA AK014582.1 - 7 - 8 1492 51
          AK046754.1 - 7 - 8 1202 51
          AK135740.1 - 7 - 8 1308 51
          AK137175.1 - 7 - 8 1309 51
          AK144323.1 - 8 - 9 1226 51
          BC036384.1 - 7 - 8 1290 1067080 51
          BC043313.1 - 2 - 3 1012 51
          BC043469.1 - 2 - 3 1012 51
          BC054826.1 - 2 - 3 1018 51
          BC094060.1 - 6 - 7 1372 51
          DQ861995.1 - 7 - 8 1156 51

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002408.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arrdc2

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954428 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954405 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954404 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954406 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957922 AK137175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957922 AK144323.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957922 AK014582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957922 AK046754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957922 AK135740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088440 AK137175.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088440 AK144323.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088440 AK014582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088440 AK046754.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088440 AK135740.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217826 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171075 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957922 AK137175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957922 AK144323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957922 AK014582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957922 AK046754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957922 AK135740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088440 AK137175.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088440 AK144323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088440 AK014582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088440 AK046754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088440 AK135740.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417651 BC094060.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417651 BC043313.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417651 BC043469.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417651 BC054826.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P417651 AK137175.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25954406 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171075 - -
          Whole Mouse Genome G4122A A_51_P417651 BC094060.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417651 BC043313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417651 BC043469.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417651 BC054826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P417651 AK137175.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171075 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06845862 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07443530 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06845860 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06845858 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954402 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954405 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954403 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954406 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954404 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25954428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25954411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954371 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06845858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06852834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07112154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06775146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07112151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07112141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06852833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06727520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06845862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07264434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25954412 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10579412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579413 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579421 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579419 - -
          Affymetrix HT MG-430 PM Array Plate 1428352_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10579406 - -
          Mouse Genome 430 2.0 Array 1428352_PM_at - -
          Mouse Genome 430 2.0 Array 1428352_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70807_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70807_at - -
          Affymetrix Mouse Clariom S Array PSR0800018347.mm.1 NM_027560.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800018347.mm.1 NM_027560.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800018347.mm.1 TC0800002408.mm.1 MTA TC
          Mouse Expression Set 430 1428352_at - -
          Mouse Expression Set 430 1428352_PM_at - -
          Murine Genome U74v2 Set 165605_PM_at - -
          Murine Genome U74v2 Set 169167_PM_at - -
          Murine Genome U74v2 Set 165605_at - -
          Murine Genome U74v2 Set 169167_at - -
          Affymetrix HT Mouse Genome 430B Array 1428352_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428352_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 350519 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104540670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2693835 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2693835 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline