Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01611736_sH
          See other OLFR169 GE Assays ›
          Gene Symbol:
          Olfr169
          Gene Name:
          olfactory receptor 169
          Gene Aliases:
          MOR273-3P
          Chromosome Location:
          Chr.16: 19565940 - 19566881 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01611736_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr169
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011855.1 NP_001011855.1 1 814 129

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr169

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123148 NM_001011855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123148 NM_001011855.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12990522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12990521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12531720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12531721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12990530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12783738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12882826 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438430 - -
          Affymetrix Mouse Gene 1.1 ST Array 10438429 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436014 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2778293 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2778293 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline