Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01612194_m1
          See other COX6A1 GE Assays ›
          Gene Symbol:
          Cox6a1
          Gene Name:
          cytochrome c oxidase subunit VIa polypeptide 1
          Gene Aliases:
          VIaL
          Chromosome Location:
          Chr.5: 115345654 - 115348955 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01612194_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cox6a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007748.3 NP_031774.1 2 - 3 259 100
          GenBank mRNA AK002402.1 - 2 - 3 261 100
          AK019318.1 - 2 - 3 264 100
          AK166496.1 - 2 - 3 261 100
          BC003807.1 - 2 - 3 259 3487598 100
          BC052816.1 - 2 - 3 265 6439987 100
          L06465.1 - 2 - 3 258 100
          U08440.1 - 2 - 3 248 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003097.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cox6a1

          Panther Classification:

          Molecular Function -

          oxidase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Process(es)

          cytochrome-c oxidase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009172 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009178 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009173 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009177 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009175 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009180 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 NM_007748.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 AK002402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 AK019318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 AK166496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 BC003807.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 NM_007748.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 AK002402.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 AK019318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 AK166496.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311540 BC003807.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48221 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 NM_007748.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 AK002402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 AK019318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 AK166496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 BC003807.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 NM_007748.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 AK002402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 AK019318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 AK166496.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311540 BC003807.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 NM_007748.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 AK002402.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 AK019318.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 AK166496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 BC003807.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 NM_007748.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 AK002402.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 AK019318.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 AK166496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P311540 BC003807.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24009158 - -
          Whole Mouse Genome G4122A A_51_P311540 NM_007748.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 AK002402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 AK019318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 AK166496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 BC003807.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 NM_007748.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 AK002402.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 AK019318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 AK166496.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311540 BC003807.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140656 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128138 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133607 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04387301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04456104 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04387300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009177 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009175 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009173 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009178 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24009172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24009146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04595473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04387402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04892093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04595466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04892094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04336839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04102585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04595468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24009168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532995 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532996 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10460150 - -
          Affymetrix HT MG-430 PM Array Plate 1417417_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532993 - -
          Affymetrix Mouse Gene 1.1 ST Array 10460149 - -
          Mouse Genome 430 2.0 Array 1417417_a_at - -
          Mouse Genome 430 2.0 Array 1417418_s_at - -
          Mouse Genome 430 2.0 Array 1417417_PM_a_at - -
          Mouse Genome 430 2.0 Array 1417418_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12861_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12861_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500027587.mm.1 TC0500003097.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027587.mm.1 NM_007748.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500027587.mm.1 NM_007748.3 Refseq
          Mouse Expression Set 430 1417418_s_at - -
          Mouse Expression Set 430 1417418_PM_s_at - -
          Mouse Expression Set 430 1417417_a_at - -
          Mouse Expression Set 430 1417417_PM_a_at - -
          Murine Genome U74v2 Set 99631_PM_f_at - -
          Murine Genome U74v2 Set 99631_f_at - -
          Mouse Genome 430A 2.0 Array 1417417_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417417_a_at - -
          Mouse Genome 430A 2.0 Array 1417418_s_at - -
          Mouse Genome 430A 2.0 Array 1417418_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417417_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417417_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106510446 - -
          Illumina mouse-6 v1.0 expression beadchip 1450239 - -
          Illumina mouse-6 v1.0 expression beadchip 1050095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001050095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2638875 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2638875 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605401 - -
          mouse-6 v1.1 (Illumina) scl0012861.1_173-S - -
          mouse-6 v1.1 (Illumina) scl012861.2_54-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450239 - -
          Illumina Sentrix mouse-6.v1 bead array 1050095 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl012861.2_54-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0012861.1_173-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1050095 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2638875 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605400 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605400 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2638875 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl012861.2_54-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0012861.1_173-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline