Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01700658_g1
          See other ZFP422 GE Assays ›
          Gene Symbol:
          Zfp422
          Gene Name:
          zinc finger protein 422
          Gene Aliases:
          2900028O21Rik, AU041373, KOX15, Krox-25-2, Krox-26, Krox25, Krox26, Znf22
          Chromosome Location:
          Chr.6: 116624016 - 116628999 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01700658_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp422
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK082970.1 - 1 - 2 48 64
          AK152681.1 - 1 - 2 32 64
          BY082381.1 - 1 - 2 70 64

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK082970.1, AK152681.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777344 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037952 AK082970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037952 NM_026057.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005465 BC018339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005465 AK003498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2005465 AK152681.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84854 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134182 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037952 AK082970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037952 NM_026057.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005465 BC018339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005465 AK003498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005465 AK152681.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P206762 AK169251.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P206762 AK003498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P206762 AK082970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P206762 AK152681.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P206762 NM_026057.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P240363 BC018339.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24777359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06038783 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134182 - -
          Whole Mouse Genome G4122A A_52_P206762 AK169251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206762 AK003498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206762 AK082970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206762 AK152681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P206762 NM_026057.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P240363 BC018339.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140968 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05283572 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05246443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24777344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24777270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05835672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05283572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05735606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05246431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05529548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05697378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05477327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05210876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05246443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24777312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05431696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547174 - -
          Affymetrix HT MG-430 PM Array Plate 1451244_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547173 - -
          Affymetrix Mouse Gene 1.1 ST Array 10449989 - -
          Mouse Genome 430 2.0 Array 1451244_PM_a_at - -
          Mouse Genome 430 2.0 Array 1451244_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67255_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67255_at - -
          Mouse Expression Set 430 1451244_a_at - -
          Mouse Expression Set 430 1451244_PM_a_at - -
          Murine Genome U74v2 Set 93350_PM_f_at - -
          Murine Genome U74v2 Set 93350_f_at - -
          Mouse Genome 430A 2.0 Array 1451244_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1451244_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451244_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451244_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 882645 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780546 - -
          Illumina mouse-6 v1.0 expression beadchip 780546 - -
          Illumina mouse-6 v1.0 expression beadchip 2320131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2435673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2477702 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2435673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2477702 - -
          mouse-6 v1.1 (Illumina) scl067255.1_28-S - -
          mouse-6 v1.1 (Illumina) scl0067255.1_297-S - -
          Illumina Sentrix mouse-6.v1 bead array 780546 - -
          Illumina Sentrix mouse-6.v1 bead array 2320131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl067255.1_28-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067255.1_297-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2435673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2477702 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2477702 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2435673 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0