Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01705384_g1
          See other HMGN2 GE Assays ›
          Gene Symbol:
          Hmgn2
          Gene Name:
          high mobility group nucleosomal binding domain 2
          Gene Aliases:
          HMG-17, Hmg17
          Chromosome Location:
          Chr.4: 133964739 - 133967991 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01705384_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmgn2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016957.3 NP_058653.1 6 1004 138
          GenBank mRNA AK002374.1 - 6 920 138
          AK078413.1 - 6 918 138
          AK146285.1 - 6 979 138
          AK146999.1 - 6 1007 138
          AK151383.1 - 6 920 138
          AK152857.1 - 6 920 138
          AK153881.1 - 1103 138
          AK169050.1 - 6 920 138
          BC083085.1 - 1091 6516041 138
          BC084680.1 - 6 861 6477991 138
          BC090836.1 - 6 904 6819800 138
          BC090837.1 - 6 894 6820123 138
          BC103792.1 - 1087 6516212 138
          X12944.1 - 865 138

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmgn2

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of transcription from RNA polymerase II promoter
          regulation of development, heterochronic

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320282 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020461 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028491 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162136 BC103792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162136 X12944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162136 BC092060.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162136 AK169050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162136 BC083085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262813 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178925 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162136 BC103792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162136 X12944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162136 BC092060.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162136 AK169050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162136 BC083085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028491 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020461 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993789 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P149864 BC103792.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P149864 X12944.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P149864 AK169050.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P149864 BC083085.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P149864 BC084680.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23320289 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178925 - -
          Whole Mouse Genome G4122A A_52_P149864 BC103792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149864 X12944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149864 AK169050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149864 BC083085.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P149864 BC084680.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102473 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105019 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106285 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178925 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03543728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03543713 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03543724 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03543707 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03543705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03543709 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320282 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23320285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23320287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03393274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03505544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03543713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04000019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23320291 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10579661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403109 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10522785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10591775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10517145 - -
          Affymetrix HT MG-430 PM Array Plate 1433507_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591773 - -
          Affymetrix Mouse Gene 1.1 ST Array 10517141 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403108 - -
          Affymetrix Mouse Gene 1.1 ST Array 10579659 - -
          Mouse Genome 430 2.0 Array 1433507_a_at - -
          Mouse Genome 430 2.0 Array 1433507_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR0400030517.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030517.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030517.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030521.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030521.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030521.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030536.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030536.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030536.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030546.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030546.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030546.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030507.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030507.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030507.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030525.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030525.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030525.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030515.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030515.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030515.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030509.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030509.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030509.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030510.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030510.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030510.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030522.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030522.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030522.mm.1 NM_016957.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030527.mm.1 TC0400003721.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400030527.mm.1 NM_016957.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400030527.mm.1 NM_016957.3 Refseq
          Mouse Expression Set 430 1433507_a_at - -
          Mouse Expression Set 430 1433507_PM_a_at - -
          Murine Genome U74v2 Set 101589_PM_at - -
          Murine Genome U74v2 Set 101589_at - -
          Mouse Genome 430A 2.0 Array 1433507_a_at - -
          Mouse Genome 430A 2.0 Array 1433507_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 893635 - -
          ABI Mouse Genome Survey Microarray 899821 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3140091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003140091 - -
          mouse-6 v1.1 (Illumina) scl015331.1_246-S - -
          Illumina Sentrix mouse-6.v1 bead array 3140091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015331.1_246-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3140091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862111 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2639081 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862111 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline