Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01952045_s1
          See other VMN1R62 GE Assays ›
          Gene Symbol:
          Vmn1r62
          Gene Name:
          vomeronasal 1 receptor 62
          Gene Aliases:
          V1rd2, V1rd8, V3R2
          Chromosome Location:
          Chr.7: 5674604 - 5676599 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01952045_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r62
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030741.2 NP_109666.2 1 1903 142
          GenBank mRNA AF324867.1 - 1 1897 142

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r62

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165989 NM_001167567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165989 BC147684.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165989 NM_030741.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165989 BC113757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2165989 BC147681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024145 NM_030741.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024145 AF324867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024145 NM_030741.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024145 AF324867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295787 AF324867.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24953322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24953310 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06115910 - -
          Whole Mouse Genome G4122A A_51_P295787 AF324867.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101651 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06115910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24953260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24953322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05926363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24953310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24953308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24953318 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10549799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10549800 - -
          Affymetrix HT MG-430 PM Array Plate 1460301_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559743 - -
          Affymetrix Mouse Gene 1.1 ST Array 10549797 - -
          Mouse Genome 430 2.0 Array 1460301_at - -
          Mouse Genome 430 2.0 Array 1460301_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81016_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81016_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700000723.mm.1 TC0700000062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000723.mm.1 NM_030741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000723.mm.1 NM_030741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000721.mm.1 TC0700000062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000721.mm.1 NM_030741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000721.mm.1 NM_030741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000722.mm.1 TC0700000062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000722.mm.1 NM_030741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000722.mm.1 NM_030741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000724.mm.1 TC0700000062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000724.mm.1 NM_030741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000724.mm.1 NM_030741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000725.mm.1 TC0700000062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000725.mm.1 NM_030741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000725.mm.1 NM_030741.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000726.mm.1 TC0700000062.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000726.mm.1 NM_030741.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000726.mm.1 NM_030741.2 Refseq
          Mouse Expression Set 430 1460301_PM_at - -
          Mouse Expression Set 430 1460301_at - -
          Mouse Genome 430A 2.0 Array 1460301_at - -
          Mouse Genome 430A 2.0 Array 1460301_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 562379 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6100278 - -
          MouseWG-6 v2 Expression BeadChip 3870451 - -
          Illumina mouse-6 v1.0 expression beadchip 6100278 - -
          Illumina mouse-6 v1.0 expression beadchip 3870451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2450137 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256964 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256964 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2450137 - -
          mouse-6 v1.1 (Illumina) scl081016.5_20-S - -
          mouse-6 v1.1 (Illumina) scl0081016.1_110-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870451 - -
          Illumina Sentrix mouse-6.v1 bead array 6100278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl081016.5_20-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0081016.1_110-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2961313 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256964 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2450137 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256964 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2961313 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2450137 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline