Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02344789_m1
          See other SMIM10L2A GE Assays ›
          Gene Symbol:
          Smim10l2a
          Gene Name:
          small integral membrane protein 10 like 2A
          Gene Aliases:
          6330419J24Rik, Ncrna00086
          Chromosome Location:
          Chr.X: 56374586 - 56378470 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02344789_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smim10l2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028086.1 1 - 2 622 111
          GenBank mRNA AK050814.1 - 1 - 2 388 111
          AK139449.1 - 1 - 2 600 111
          BC052359.1 - 1 - 2 593 6585408 111
          BQ444292.1 - 1 - 2 622 111

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000516.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958583 BC052359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958583 AK078069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958583 AK139449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958583 NR_028086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958583 AK050814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 BC052359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 AK078069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 AK139449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 NR_028086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 AK050814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 BC052359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 AK078069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 AK139449.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 NR_028086.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P247614 AK050814.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814661 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176461 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147540 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P176461 - -
          Whole Mouse Genome G4122A A_51_P247614 BC052359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 AK078069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 AK139449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 NR_028086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 AK050814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 BC052359.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 AK078069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 AK139449.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 NR_028086.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P247614 AK050814.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P176461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14656852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32395582 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599694 - -
          Affymetrix HT MG-430 PM Array Plate 1435200_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10599693 - -
          Mouse Genome 430 2.0 Array 1435200_at - -
          Mouse Genome 430 2.0 Array 1435200_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00003373.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003373.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003373.mm.1 TC0X00000516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003378.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003378.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003378.mm.1 TC0X00000516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003372.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003372.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003372.mm.1 TC0X00000516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003374.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003374.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003374.mm.1 TC0X00000516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003370.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003370.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003370.mm.1 TC0X00000516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003375.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003375.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003375.mm.1 TC0X00000516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003371.mm.1 NR_028086.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003371.mm.1 NR_028086.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003371.mm.1 TC0X00000516.mm.1 MTA TC
          Mouse Expression Set 430 1435200_at - -
          Mouse Expression Set 430 1435200_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435200_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435200_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 897257 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline