Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02345576_s1
          See other OLFR901 GE Assays ›
          Gene Symbol:
          Olfr901
          Gene Name:
          olfactory receptor 901
          Gene Aliases:
          MOR162-8
          Chromosome Location:
          Chr.9: 38429455 - 38431219 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02345576_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr901
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011806.1 NP_001011806.1 1 757 139

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr901

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045581 NM_001011806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045581 NM_001011806.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26434656 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07525768 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08059455 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07525774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07525770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08059450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26434691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07525774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07498049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08059444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26434668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07525772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584408 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584407 - -
          Affymetrix Mouse Clariom S Array PSR0900002798.mm.1 NM_001011806.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002798.mm.1 NM_001011806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900002798.mm.1 TC0900000399.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162504 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162504 - -
          mouse-6 v1.1 (Illumina) GI_58801377-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162504 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162504 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline