Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02376605_s1
          See other 9130230L23RIK GE Assays ›
          Gene Symbol:
          9130230L23Rik
          Gene Name:
          RIKEN cDNA 9130230L23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 65987928 - 66004285 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02376605_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9130230L23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027961.1 7 1345 132
          GenBank mRNA AK033717.1 - 7 1346 132

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P206165 BC096478.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P206165 NR_027961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P206165 AK033717.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P206165 BC096478.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P206165 NR_027961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P206165 AK033717.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647711 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678678 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 526907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 321268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78616 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143398 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P206165 BC096478.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P206165 NR_027961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P206165 AK033717.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P206165 BC096478.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P206165 NR_027961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P206165 AK033717.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01121351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143398 - -
          Whole Mouse Genome G4122A A_52_P674348 AK078705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206165 BC096478.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206165 NR_027961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206165 AK033717.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206165 BC096478.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206165 NR_027961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P206165 AK033717.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04740556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04103581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04521388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04372826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04180782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04223164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04688620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04180784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04858160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04446066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04353812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04300466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04715394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04714758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04758930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04180785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04617382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04688623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04525551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04353811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04866093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23759372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04180787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04929140 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10530262 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530265 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530266 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530263 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530264 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530261 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530267 - -
          Affymetrix Mouse Gene 1.0 ST Array 10530260 - -
          Affymetrix Mouse Gene 1.1 ST Array 10530259 - -
          Affymetrix Mouse Clariom S Array PSR0500022310.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022310.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022310.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022315.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022315.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022315.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022326.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022326.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022326.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022312.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022312.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022312.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022311.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022311.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022311.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022321.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022321.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022321.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022323.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022323.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022323.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022318.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022318.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022318.mm.1 NR_027961.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022324.mm.1 TC0500002503.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022324.mm.1 NR_027961.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022324.mm.1 NR_027961.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 833238 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline