Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02391637_s1
          See other OLFR917 GE Assays ›
          Gene Symbol:
          Olfr917
          Gene Name:
          olfactory receptor 917
          Gene Aliases:
          MOR168-2P
          Chromosome Location:
          Chr.9: 38664913 - 38665842 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02391637_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr917
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011864.1 NP_001011864.1 1 661 133

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr917

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1952046 NM_001011864.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1952046 NM_001011864.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435559 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07762239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07875813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07875796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08171388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07875814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07875810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07792555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07762235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592377 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592376 - -
          Affymetrix Mouse Clariom S Array PSR0900017174.mm.1 NM_001011864.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017174.mm.1 NM_001011864.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017174.mm.1 TC0900002062.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2930554 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2930554 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0