Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02524882_s1
          See other DPM3 GE Assays ›
          Gene Symbol:
          Dpm3
          Gene Name:
          dolichyl-phosphate mannosyltransferase polypeptide 3
          Gene Aliases:
          1110001H19Rik, AV117483
          Chromosome Location:
          Chr.3: 89259263 - 89267079 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02524882_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpm3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026767.4 NP_081043.1 2 424 80
          GenBank mRNA AI527477.1 - 1 368 80
          AK003223.1 - 2 334 80
          BC037761.1 - 1 368 80
          CJ237809.1 - 2 424 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dpm3

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          regulation of protein stability
          protein O-linked mannosylation
          dolichyl-phosphate beta-D-mannosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318234 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318232 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010768 NM_026767.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010768 AK003223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010768 BC037761.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145175 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010768 NM_026767.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010768 AK003223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010768 BC037761.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22318242 - -
          Whole Mouse Genome G4122A A_51_P518558 NM_026767.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P518558 AK003223.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03034590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03034595 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03034589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02644034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318233 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22318234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22318238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03034595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03034589 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22318222 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493492 - -
          Affymetrix HT MG-430 PM Array Plate 1452729_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493490 - -
          Mouse Genome 430 2.0 Array 1452729_PM_at - -
          Mouse Genome 430 2.0 Array 1452729_at - -
          Mouse Expression Set 430 1452729_at - -
          Mouse Expression Set 430 1452729_PM_at - -
          Murine Genome U74v2 Set 101084_PM_f_at - -
          Murine Genome U74v2 Set 101084_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1452729_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452729_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 453149 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715803 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715803 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline