Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02526026_g1
          See other 2410018L13RIK GE Assays ›
          Gene Symbol:
          2410018L13Rik
          Gene Name:
          RIKEN cDNA 2410018L13 gene
          Gene Aliases:
          2810480C08Rik
          Chromosome Location:
          Chr.12: 22953997 - 23010243 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02526026_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2410018L13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC063063.1 - 4 2471 179

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068406 BC063063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068406 NR_015504.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068406 AK134640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976470 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068406 BC063063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068406 NR_015504.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068406 AK134640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976470 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10394806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394834 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394854 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394841 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394838 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394840 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394835 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394836 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399674 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399645 - -
          Affymetrix HT MG-430 PM Array Plate 1431317_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430845_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10394940 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394934 - -
          Affymetrix Mouse Gene 1.1 ST Array 10399659 - -
          Affymetrix Mouse Gene 1.1 ST Array 10399655 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394837 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394827 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394931 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394852 - -
          Affymetrix Mouse Gene 1.1 ST Array 10399640 - -
          Mouse Genome 430 2.0 Array 1430845_PM_at - -
          Mouse Genome 430 2.0 Array 1431317_PM_at - -
          Mouse Genome 430 2.0 Array 1430845_at - -
          Mouse Genome 430 2.0 Array 1431317_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69732_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69732_PM_at - -
          Mouse Expression Set 430 1439113_at - -
          Mouse Expression Set 430 1439113_PM_at - -
          Mouse Expression Set 430 1430845_PM_at - -
          Mouse Expression Set 430 1430845_at - -
          Murine Genome U74v2 Set 104641_f_at - -
          Murine Genome U74v2 Set 104641_PM_f_at - -
          Murine Genome U74v2 Set 139031_PM_f_at - -
          Murine Genome U74v2 Set 139031_f_at - -
          Mouse Genome 430A 2.0 Array 1430845_at - -
          Mouse Genome 430A 2.0 Array 1439113_PM_at - -
          Mouse Genome 430A 2.0 Array 1430845_PM_at - -
          Mouse Genome 430A 2.0 Array 1439113_at - -
          Affymetrix HT Mouse Genome 430A Array 1430845_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1430845_at - -
          Affymetrix HT Mouse Genome 430B Array 1431317_at - -
          Affymetrix HT Mouse Genome 430B Array 1431317_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738005 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738005 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2542320 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2738005 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738005 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline