Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02528467_g1
          See other RPL32 GE Assays ›
          Gene Symbol:
          Rpl32
          Gene Name:
          ribosomal protein L32
          Gene Aliases:
          AU020185, rpL32-3A
          Chromosome Location:
          Chr.6: 115805514 - 115808743 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02528467_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172086.2 NP_742083.1 4 435 69
          GenBank mRNA AK002353.1 - 4 436 69
          AK011017.1 - 4 436 69
          AK012525.1 - 4 436 69
          AK028204.1 - 4 436 69
          AK146768.1 - 4 436 69
          AK148453.1 - 4 436 69
          AK150705.1 - 4 436 69
          AK168282.1 - 4 439 69
          BC030176.1 - 434 69
          BC046339.1 - 4 404 6478080 69

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl32

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773075 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773079 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773080 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773076 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773077 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149049 BC030176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149049 BC046339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149049 AK002353.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149049 NM_172086.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2149049 AK011017.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410426 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105026 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149049 BC030176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149049 BC046339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149049 AK002353.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149049 NM_172086.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2149049 AK011017.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P573161 BC030176.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P573161 BC046339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P573161 BC086909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P573161 AK002353.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P573161 NM_172086.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24773056 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105026 - -
          Whole Mouse Genome G4122A A_52_P573161 BC030176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P573161 BC046339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P573161 BC086909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P573161 AK002353.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P573161 NM_172086.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123934 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105026 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557502 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05154144 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557504 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557505 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557495 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557496 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05557498 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773076 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773080 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773075 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24773079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24773059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05153396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24773080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05153395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05153402 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547072 - -
          Affymetrix HT MG-430 PM Array Plate 1435897_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547068 - -
          Mouse Genome 430 2.0 Array 1435897_at - -
          Mouse Genome 430 2.0 Array 1435897_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600022746.mm.1 NM_172086.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022746.mm.1 NM_172086.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022746.mm.1 TC0600002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022744.mm.1 NM_172086.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022744.mm.1 NM_172086.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022744.mm.1 TC0600002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022745.mm.1 NM_172086.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022745.mm.1 NM_172086.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022745.mm.1 TC0600002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600022748.mm.1 NM_172086.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022748.mm.1 NM_172086.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600022748.mm.1 TC0600002960.mm.1 MTA TC
          Mouse Expression Set 430 1435897_PM_at - -
          Mouse Expression Set 430 1435897_at - -
          Murine Genome U74v2 Set 160478_PM_r_at - -
          Murine Genome U74v2 Set 160478_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1435897_at - -
          Affymetrix HT Mouse Genome 430B Array 1435897_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895735 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 840341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754337 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754337 - -
          mouse-6 v1.1 (Illumina) scl019951.1_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 840341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl019951.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840341 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232890 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl019951.1_25-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5cvsx:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline