Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02600024_m1
          See other GM156 GE Assays ›
          Gene Symbol:
          Gm156
          Gene Name:
          predicted gene 156
          Gene Aliases:
          Klrh1
          Chromosome Location:
          Chr.6: 129766568 - 129784270 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02600024_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm156
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014997.1 NP_001014997.1 5 - 6 547 67
          GenBank mRNA AB121775.1 - 5 - 6 547 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003248.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003306 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127627 AK138706.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118308 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003306 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127627 AK138706.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06068795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06068811 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06068781 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06068795 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06068811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24845375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845400 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05692100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05740389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05107373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24845436 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548436 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548430 - -
          Affymetrix Mouse Clariom S Array PSR0600025386.mm.1 TC0600003248.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025386.mm.1 NM_001014997.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025386.mm.1 NM_001014997.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258224 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2987890 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2987890 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline