Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02600578_m1
          See other LRRC55 GE Assays ›
          Gene Symbol:
          Lrrc55
          Gene Name:
          leucine rich repeat containing 55
          Gene Aliases:
          A330097E02Rik, Gm351
          Chromosome Location:
          Chr.2: 85188067 - 85198932 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02600578_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc55
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033346.2 NP_001028518.1 1 - 2 721 119
          GenBank mRNA AK134968.1 - 722 119
          BC141221.1 - 721 9055833 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003888.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2058711 AK134968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P395280 AK086116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P395280 NM_001033346.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P395280 AK039730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P395280 AK086116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P395280 NM_001033346.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P395280 AK039730.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673609 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94826 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167658 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P395280 AK086116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P395280 NM_001033346.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P395280 AK039730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P395280 AK086116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P395280 NM_001033346.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P395280 AK039730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2058711 AK134968.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00423354 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04592385 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167658 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04592385 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21396829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01564667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02215646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01524350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02162178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01410019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02046117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02231765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02043308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01672306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02046122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02229985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02043309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01420610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02245647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02044389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01436547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01768523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01672304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02215647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02231763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01169640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02231759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01410027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396821 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01286321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01524349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02229986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01254015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01298083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02130913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01672305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01524344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01299511 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21396707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01794450 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10484504 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484503 - -
          Affymetrix Mouse Clariom S Array PSR0200033662.mm.1 NM_001033346.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033662.mm.1 NM_001033346.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033662.mm.1 TC0200003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033666.mm.1 NM_001033346.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033666.mm.1 NM_001033346.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033666.mm.1 TC0200003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033659.mm.1 TC0200003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033663.mm.1 NM_001033346.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033663.mm.1 NM_001033346.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033663.mm.1 TC0200003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033664.mm.1 NM_001033346.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033664.mm.1 NM_001033346.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033664.mm.1 TC0200003888.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200033667.mm.1 NM_001033346.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033667.mm.1 NM_001033346.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033667.mm.1 TC0200003888.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 517006 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257870 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215600 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline