Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02602271_mH
          See other CYP2C54 GE Assays ›
          Gene Symbol:
          Cyp2c54
          Gene Name:
          cytochrome P450, family 2, subfamily c, polypeptide 54
          Gene Aliases:
          EG404195
          Chromosome Location:
          Chr.19: 40037940 - 40073845 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02602271_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2c54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206537.2 NP_996260.1 5 - 6 819 109
          GenBank mRNA AI047349.1 - 2 - 3 255 109
          BC043929.1 - 4 - 5 601 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001481.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2c54

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          arachidonic acid metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          linoleic acid metabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding
          metal ion binding
          aromatase activity
          linoleic acid epoxygenase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P154580 NM_206537.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P154580 AI047349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P154580 BC043929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P154580 NM_206537.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P154580 AI047349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P154580 BC043929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993419 NM_206537.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993419 AI046359.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1993419 BC043929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158547 NM_206537.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158547 AI047349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158547 BC043929.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783688 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692219 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126588 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142331 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118314 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113218 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163617 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165574 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140803 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125465 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P154580 NM_206537.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P154580 AI047349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P154580 BC043929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P154580 NM_206537.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P154580 AI047349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P154580 BC043929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158547 NM_206537.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158547 AI047349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158547 BC043929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993419 NM_206537.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993419 AI046359.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1993419 BC043929.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32106966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32106967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32106965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32106964 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09170061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169951 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165574 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140803 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169937 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125465 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113218 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169999 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09169921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09170024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09170015 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118314 - -
          Whole Mouse Genome G4122A A_52_P154580 NM_206537.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154580 AI047349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154580 BC043929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154580 NM_206537.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154580 AI047349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P154580 BC043929.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165574 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113218 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163617 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125465 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140803 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118314 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09169907 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14486635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32106938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32106964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10467401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10467406 - -
          Affymetrix HT MG-430 PM Array Plate 1455457_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10467400 - -
          Mouse Genome 430 2.0 Array 1455457_PM_at - -
          Mouse Genome 430 2.0 Array 1455457_at - -
          Affymetrix Mouse Clariom S Array PSR1900012929.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012929.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012929.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012931.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012931.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012931.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012924.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012924.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012924.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012925.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012925.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012925.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012932.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012932.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012932.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012933.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012933.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012933.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012927.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012927.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012927.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012926.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012926.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012926.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012928.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012928.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012928.mm.1 TC1900001481.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900012930.mm.1 NM_206537.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012930.mm.1 NM_206537.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900012930.mm.1 TC1900001481.mm.1 MTA TC
          Mouse Expression Set 430 1455457_at - -
          Mouse Expression Set 430 1455457_PM_at - -
          Mouse Genome 430A 2.0 Array 1455457_PM_at - -
          Mouse Genome 430A 2.0 Array 1455457_at - -
          Affymetrix HT Mouse Genome 430A Array 1455457_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455457_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 704791 - -
          ABI Mouse Genome Survey Microarray 920848 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6130193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680726 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2642175 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2642175 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2680726 - -
          mouse-6 v1.1 (Illumina) scl000511.1_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000511.1_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130193 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241818 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline