Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02745310_gH
          See other ZFP953 GE Assays ›
          Gene Symbol:
          Zfp953
          Gene Name:
          zinc finger protein 953
          Gene Aliases:
          E130120F12Rik
          Chromosome Location:
          Chr.13: 67339309 - 67360572 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02745310_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp953
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001038651.3 NP_001033740.1 3 - 4 290 70
          GenBank mRNA AK143165.1 - 3 - 4 291 70
          BC141233.1 - 3 - 4 266 9055845 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002773.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045050 NM_001038651.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045050 AK143165.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 575500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864827 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773785 AK085850.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 129997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 47122 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166227 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102145 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122974 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045050 NM_001038651.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045050 AK143165.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102145 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07905520 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122974 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166227 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102145 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29108244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10520166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11076448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11076467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11076455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108293 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29108298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410307 - -
          Affymetrix HT MG-430 PM Array Plate 1439176_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10410302 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 629016_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 629016_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300018651.mm.1 NM_001038651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018651.mm.1 NM_001038651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018651.mm.1 TC1300002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018650.mm.1 NM_001038651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018650.mm.1 NM_001038651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018650.mm.1 TC1300002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018652.mm.1 NM_001038651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018652.mm.1 NM_001038651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018652.mm.1 TC1300002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014599.mm.1 NM_001038651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014599.mm.1 NM_001038651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014599.mm.1 TC1300002773.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014603.mm.1 NM_001038651.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014603.mm.1 NM_001038651.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014603.mm.1 TC1300002773.mm.1 MTA TC
          Mouse Expression Set 430 1439176_PM_at - -
          Mouse Expression Set 430 1439176_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220082 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline