Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02765926_s1
          See other ZFP873 GE Assays ›
          Gene Symbol:
          Zfp873
          Gene Name:
          zinc finger protein 873
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 82048086 - 82077727 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02765926_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp873
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024626.2 NP_001019797.2 3 338 143
          GenBank mRNA AK143655.1 - 3 343 143
          BC062115.1 - 1 365 143
          BC090848.1 - 3 322 30626866 143
          BC096521.1 - 2 132 143

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291405 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291409 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291406 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291278 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975852 BC090848.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975852 BC096521.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975852 NM_001024626.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975852 AK143655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1975852 BC062115.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 95232 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172840 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975852 BC090848.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975852 BC096521.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975852 NM_001024626.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975852 AK143655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1975852 BC062115.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27291200 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07126342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172840 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08221679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08738772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291405 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291280 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291409 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27291406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27291181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08840797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08672419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27291182 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10365210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365209 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10365215 - -
          Affymetrix HT MG-430 PM Array Plate 1446919_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10365208 - -
          Mouse Genome 430 2.0 Array 1446919_PM_at - -
          Mouse Genome 430 2.0 Array 1446919_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 408062_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 408062_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000007319.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007319.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007319.mm.1 TC1000000915.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007320.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007320.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007320.mm.1 TC1000000915.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007318.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007318.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007318.mm.1 TC1000000915.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007322.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007322.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007322.mm.1 TC1000000915.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007323.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007323.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007323.mm.1 TC1000000915.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007321.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007321.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007321.mm.1 TC1000000915.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007324.mm.1 NM_001024626.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007324.mm.1 NM_001024626.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007324.mm.1 TC1000000915.mm.1 MTA TC
          Mouse Expression Set 430 1446919_PM_at - -
          Mouse Expression Set 430 1446919_at - -
          Murine Genome U74v2 Set 130584_at - -
          Murine Genome U74v2 Set 130584_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446919_at - -
          Affymetrix HT Mouse Genome 430B Array 1446919_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241808 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2913869 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913869 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline