Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015167_s1
          See other GM5346 GE Assays ›
          Gene Symbol:
          Gm5346
          Gene Name:
          predicted gene 5346
          Gene Aliases:
          EG384813
          Chromosome Location:
          Chr.8: 43624957 - 43703316 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015167_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5346
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025240.1 NP_001020411.1 1 943 144
          GenBank mRNA BC151154.1 - 1 1035 9087881 144
          BC157897.1 - 1 1035 9087877 144

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gm5346

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013725 BC157897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013725 NM_001025240.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013725 BC151154.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14340 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013725 BC157897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013725 NM_001025240.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013725 BC151154.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25821005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25821013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25820991 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06493716 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06493716 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07419873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25821013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25820991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25821012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06741415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25821005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25820979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25821016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25821013 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10578422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578414 - -
          Affymetrix Mouse Gene 1.1 ST Array 10578421 - -
          Affymetrix Mouse Gene 1.1 ST Array 10578413 - -
          Affymetrix Mouse Clariom S Array PSR0800016412.mm.1 TC0800002133.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800016410.mm.1 NM_001025240.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016410.mm.1 NM_001025240.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016410.mm.1 TC0800002133.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 432818 - -
          ABI Mouse Genome Survey Microarray 854253 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622239 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2622239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3056397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3133209 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3133209 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3056397 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline