Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03020030_g1
          See other RPL23A GE Assays ›
          Gene Symbol:
          Rpl23a
          Gene Name:
          ribosomal protein L23A
          Gene Aliases:
          BC029892, MDA20
          Chromosome Location:
          Chr.11: 78180935 - 78183584 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03020030_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl23a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207523.2 NP_997406.1 2 84 125
          GenBank mRNA AF452721.1 - 2 43 125
          AK028387.1 - 2 57 125
          AK152095.1 - 2 57 125
          BC026656.1 - 2 41 125
          BC029892.1 - 2 43 4163533 125
          BC086883.1 - 2 59 6756980 125
          BC086884.1 - 2 57 30122733 125
          BC096637.1 - 2 42 125
          BC125640.1 - 2 63 40130155 125
          BC125642.1 - 2 63 40130157 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl23a

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          nucleotide binding
          RNA binding
          structural constituent of ribosome
          protein binding
          rRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941829 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941846 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109874 BC029892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109874 BC086883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109874 BC086884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109874 BC096637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109874 BC125640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 581200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218386 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78998 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120285 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109875 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109874 BC029892.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109874 BC086883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109874 BC086884.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109874 BC096637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109874 BC125640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P685714 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127516 BC029892.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127516 BC086883.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127516 BC086884.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127516 BC096637.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P127516 BC026656.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941842 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120285 - -
          Whole Mouse Genome G4122A A_52_P685714 - -
          Whole Mouse Genome G4122A A_51_P127516 BC029892.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127516 BC086883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127516 BC086884.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127516 BC096637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P127516 BC026656.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106666 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120285 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09663267 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09663268 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09663278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09295588 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09295595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941829 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09457744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08978078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09295588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09246563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941832 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421708 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371266 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371270 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388779 - -
          Affymetrix Mouse Gene 1.0 ST Array 10579658 - -
          Affymetrix Mouse Gene 1.0 ST Array 10346106 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371268 - -
          Affymetrix Mouse Gene 1.0 ST Array 10350991 - -
          Affymetrix Mouse Gene 1.0 ST Array 10361817 - -
          Affymetrix HT MG-430 PM Array Plate 1426660_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371269 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598831 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598103 - -
          Affymetrix Mouse Gene 1.1 ST Array 10579657 - -
          Affymetrix Mouse Gene 1.1 ST Array 10350990 - -
          Affymetrix Mouse Gene 1.1 ST Array 10371265 - -
          Affymetrix Mouse Gene 1.1 ST Array 10414078 - -
          Affymetrix Mouse Gene 1.1 ST Array 10346105 - -
          Affymetrix Mouse Gene 1.1 ST Array 10421707 - -
          Affymetrix Mouse Gene 1.1 ST Array 10361816 - -
          Affymetrix Mouse Gene 1.1 ST Array 10371267 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388776 - -
          Affymetrix Mouse Gene 1.1 ST Array 10371275 - -
          Mouse Genome 430 2.0 Array 1438927_x_at - -
          Mouse Genome 430 2.0 Array 1437975_a_at - -
          Mouse Genome 430 2.0 Array 1437976_x_at - -
          Mouse Genome 430 2.0 Array 1426660_x_at - -
          Mouse Genome 430 2.0 Array 1438927_PM_x_at - -
          Mouse Genome 430 2.0 Array 1426659_PM_a_at - -
          Mouse Genome 430 2.0 Array 1437976_PM_x_at - -
          Mouse Genome 430 2.0 Array 1426659_a_at - -
          Mouse Genome 430 2.0 Array 1437975_PM_a_at - -
          Mouse Genome 430 2.0 Array 1426660_PM_x_at - -
          Affymetrix Mouse Clariom S Array PSR1100030278.mm.1 NM_207523.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030278.mm.1 NM_207523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030278.mm.1 TC1100003288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030280.mm.1 NM_207523.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030280.mm.1 NM_207523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030280.mm.1 TC1100003288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100030281.mm.1 NM_207523.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030281.mm.1 NM_207523.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100030281.mm.1 TC1100003288.mm.1 MTA TC
          Mouse Expression Set 430 1438927_x_at - -
          Mouse Expression Set 430 1426660_PM_x_at - -
          Mouse Expression Set 430 1426660_x_at - -
          Mouse Expression Set 430 1438927_PM_x_at - -
          Mouse Expression Set 430 1437976_x_at - -
          Mouse Expression Set 430 1437976_PM_x_at - -
          Mouse Expression Set 430 1426659_a_at - -
          Mouse Expression Set 430 1426659_PM_a_at - -
          Mouse Expression Set 430 1437975_a_at - -
          Mouse Expression Set 430 1437975_PM_a_at - -
          Murine Genome U74v2 Set 94823_PM_at - -
          Murine Genome U74v2 Set 94823_at - -
          Mouse Genome 430A 2.0 Array 1438927_x_at - -
          Mouse Genome 430A 2.0 Array 1437976_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1426660_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437976_x_at - -
          Mouse Genome 430A 2.0 Array 1426659_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426659_a_at - -
          Mouse Genome 430A 2.0 Array 1426660_x_at - -
          Mouse Genome 430A 2.0 Array 1437975_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1438927_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1437975_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 894181 - -
          ABI Mouse Genome Survey Microarray 896878 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1450403 - -
          Illumina mouse-6 v1.0 expression beadchip 1450403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236631 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236631 - -
          mouse-6 v1.1 (Illumina) scl0268449.4_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0268449.4_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236631 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236631 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0268449.4_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline