Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03030414_g1
          See other RPL36 GE Assays ›
          Gene Symbol:
          Rpl36
          Gene Name:
          ribosomal protein L36
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 56613395 - 56614246 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03030414_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rpl36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_018730.3 NP_061200.2 3 200 121
          GenBank mRNA AK003115.1 - 3 180 121
          BC021595.1 - 3 168 5100307 121
          BC086914.1 - 3 179 6768133 121
          BY055319.1 - 3 202 121
          CF728650.1 - 2 277 121
          X75895.1 - 2 164 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rpl36

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          molecular_function
          structural constituent of ribosome

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257292 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976983 CF728650.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976983 NM_018730.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976983 BC021595.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976983 BC086914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002113 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33256 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101786 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976983 CF728650.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976983 NM_018730.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976983 BC021595.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976983 BC086914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002113 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071326 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P361875 X75895.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P233312 NM_001195765.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P233312 CF728650.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P233312 NM_018730.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P233312 AK003115.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P233312 BC021595.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257289 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101786 - -
          Whole Mouse Genome G4122A A_51_P361875 X75895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P233312 NM_001195765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P233312 CF728650.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P233312 NM_018730.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P233312 AK003115.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P233312 BC021595.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130460 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108495 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13290090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13338959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13290092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10594947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10394610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10378784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10426649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10592139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446133 - -
          Affymetrix Mouse Gene 1.1 ST Array 10546801 - -
          Affymetrix Mouse Gene 1.1 ST Array 10446131 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378783 - -
          Affymetrix Mouse Gene 1.1 ST Array 10394609 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471358 - -
          Affymetrix Mouse Gene 1.1 ST Array 10426648 - -
          Affymetrix Mouse Gene 1.1 ST Array 10515383 - -
          Affymetrix Mouse Gene 1.1 ST Array 10431912 - -
          Affymetrix Mouse Gene 1.1 ST Array 10594946 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592138 - -
          Mouse Genome 430 2.0 Array 1433510_x_at - -
          Mouse Genome 430 2.0 Array 1416519_PM_at - -
          Mouse Genome 430 2.0 Array 1416520_PM_x_at - -
          Mouse Genome 430 2.0 Array 1416520_x_at - -
          Mouse Genome 430 2.0 Array 1433510_PM_x_at - -
          Mouse Genome 430 2.0 Array 1416519_at - -
          Affymetrix Mouse Clariom S Array PSR1700009723.mm.1 NM_018730.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009723.mm.1 NM_018730.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009723.mm.1 TC1700001012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009725.mm.1 NM_018730.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009725.mm.1 NM_018730.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009725.mm.1 TC1700001012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700009724.mm.1 NM_018730.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009724.mm.1 NM_018730.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009724.mm.1 TC1700001012.mm.1 MTA TC
          Mouse Expression Set 430 1433510_PM_x_at - -
          Mouse Expression Set 430 1433510_x_at - -
          Mouse Expression Set 430 1416519_PM_at - -
          Mouse Expression Set 430 1416519_at - -
          Mouse Expression Set 430 1416520_PM_x_at - -
          Mouse Expression Set 430 1416520_x_at - -
          Murine Genome U74v2 Set 92628_at - -
          Murine Genome U74v2 Set 92628_PM_at - -
          Mouse Genome 430A 2.0 Array 1416519_at - -
          Mouse Genome 430A 2.0 Array 1416520_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1433510_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1433510_x_at - -
          Mouse Genome 430A 2.0 Array 1416519_PM_at - -
          Mouse Genome 430A 2.0 Array 1416520_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895253 - -
          ABI Mouse Genome Survey Microarray 900234 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6450347 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006450347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232274 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232274 - -
          mouse-6 v1.1 (Illumina) scl054217.2_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 6450347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl054217.2_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6450347 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232274 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl054217.2_13-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline