Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03048204_m1
          See other SCGB1B27 GE Assays ›
          Gene Symbol:
          Scgb1b27
          Gene Name:
          secretoglobin, family 1B, member 27
          Gene Aliases:
          Abp, Abpa, Abpa27, Sal-1, Tcp
          Chromosome Location:
          Chr.7: 34021567 - 34022881 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03048204_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scgb1b27
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009596.1 NP_033726.1 2 - 3 245 109
          GenBank mRNA BC016132.1 - 2 - 3 245 4922791 109
          BC132436.1 - 2 - 3 329 40130713 109

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000628.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scgb1b27

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          steroid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090725 NM_009596.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090725 BC016132.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090725 BC132436.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090725 GU269231.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090725 NM_009596.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090725 BC016132.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090725 BC132436.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090725 GU269231.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344136 AF039064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P87703 NM_009596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P87703 BC016132.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P87703 BC132436.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P87703 GU269231.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552122 - -
          Affymetrix HT MG-430 PM Array Plate 1424063_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552121 - -
          Mouse Genome 430 2.0 Array 1424063_at - -
          Mouse Genome 430 2.0 Array 1424063_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11354_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11354_at - -
          Affymetrix Mouse Clariom S Array PSR0700005369.mm.1 TC0700000628.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005369.mm.1 NM_009596.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005369.mm.1 NM_009596.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005370.mm.1 TC0700000628.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700005370.mm.1 NM_009596.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700005370.mm.1 NM_009596.1 Refseq
          Mouse Expression Set 430 1424063_PM_at - -
          Mouse Expression Set 430 1424063_at - -
          Murine Genome U74v2 Set 94628_r_at - -
          Murine Genome U74v2 Set 94628_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1424063_at - -
          Mouse Genome 430A 2.0 Array 1424063_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424063_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424063_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5050270 - -
          Illumina mouse-6 v1.0 expression beadchip 5050270 - -
          Illumina mouse-6 v1.0 expression beadchip 2360048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050270 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249191 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703052 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703052 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249191 - -
          mouse-6 v1.1 (Illumina) scl011354.2_35-S - -
          mouse-6 v1.1 (Illumina) scl0011354.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050270 - -
          Illumina Sentrix mouse-6.v1 bead array 2360048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl011354.2_35-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0011354.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050270 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2703052 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249191 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2703052 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249191 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline