Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03053975_g1
          See other ZFP42 GE Assays ›
          Gene Symbol:
          Zfp42
          Gene Name:
          zinc finger protein 42
          Gene Aliases:
          Rex-1, Rex1, Zfp-42
          Chromosome Location:
          Chr.8: 43295067 - 43307009 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03053975_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp42
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009556.3 NP_033582.2 4 1082 111
          GenBank mRNA AB221555.1 - 1 681 111
          BC053399.1 - 4 1055 30031349 111
          BC064785.1 - 5 1113 111
          BC098467.1 - 4 1017 30008039 111
          M28382.1 - 4 1043 111

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp42

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          spermatid development
          regulation of genetic imprinting
          nucleic acid binding
          DNA binding
          protein binding
          sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413376 BC098467.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P413376 BC098467.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 261581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174349 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134977 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139210 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413376 BC098467.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P413376 BC098467.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413376 BC098467.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P413376 BC098467.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25819779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06493139 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134977 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139210 - -
          Whole Mouse Genome G4122A A_51_P413376 BC098467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P413376 BC098467.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629849 M28382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P629849 BC098467.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139911 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134977 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07445222 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440662 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07445221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07445225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25819741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06912286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07366915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06937510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06785463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07445226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06912294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06912292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06761482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07028536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06810171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07026653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06684827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06703546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25819795 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10578404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578403 - -
          Affymetrix HT MG-430 PM Array Plate 1418362_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10578401 - -
          Mouse Genome 430 2.0 Array 1418362_at - -
          Mouse Genome 430 2.0 Array 1418362_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22702_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22702_PM_at - -
          Mouse Expression Set 430 1418362_PM_at - -
          Mouse Expression Set 430 1418362_at - -
          Murine Genome U74v2 Set 98414_PM_at - -
          Murine Genome U74v2 Set 98414_at - -
          Mouse Genome 430A 2.0 Array 1418362_at - -
          Mouse Genome 430A 2.0 Array 1418362_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418362_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418362_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 916512 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1340402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001340402 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220578 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220578 - -
          mouse-6 v1.1 (Illumina) scl022702.4_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 1340402 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl022702.4_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1340402 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2961604 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2961604 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline