Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03647531_s1
          See other GM7714 GE Assays ›
          Gene Symbol:
          Gm7714
          Gene Name:
          predicted gene 7714
          Gene Aliases:
          EG665615
          Chromosome Location:
          Chr.5: 88268919 - 88282835 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03647531_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm7714
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110779.1 NP_001104249.1 3 354 126
          GenBank mRNA AK137735.1 - 3 356 126

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062078 NM_001110779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062078 AK137735.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122553 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158079 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100116 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062078 NM_001110779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062078 AK137735.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100116 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154869 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646281 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646384 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158079 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154853 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646372 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646402 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646442 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154846 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154830 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646304 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01154886 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646346 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05646336 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01154886 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05646442 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01154853 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158079 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01154846 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01154830 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23871630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04313094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04316141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23871634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500007091.mm.1 TC0500000808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007091.mm.1 NM_001110779.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007091.mm.1 NM_001110779.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007092.mm.1 TC0500000808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007092.mm.1 NM_001110779.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007092.mm.1 NM_001110779.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007093.mm.1 TC0500000808.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007093.mm.1 NM_001110779.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007093.mm.1 NM_001110779.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0