Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03647545_s1
          See other VMN2R10 GE Assays ›
          Gene Symbol:
          Vmn2r10
          Gene Name:
          vomeronasal 2, receptor 10
          Gene Aliases:
          V2r16, VR16
          Chromosome Location:
          Chr.5: 108995539 - 109006436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03647545_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009491.3 NP_033517.3 2 342 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn2r10

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2012107 NM_009491.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P216576 AF011426.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P216576 AF011426.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142291 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2012107 NM_009491.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P216576 AF011426.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P216576 AF011426.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23974854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23974840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23974836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23974845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23974863 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05690960 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142291 - -
          Whole Mouse Genome G4122A A_51_P216576 AF011426.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P216576 AF011426.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135634 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142291 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05690960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04919047 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04919031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04919028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23974848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04953951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04649992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04919035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04582805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04582797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04919047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23974872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04582803 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532272 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532273 - -
          Affymetrix HT MG-430 PM Array Plate 1421701_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532270 - -
          Mouse Genome 430 2.0 Array 1421701_PM_at - -
          Mouse Genome 430 2.0 Array 1421701_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22307_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22307_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500026215.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026215.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026215.mm.1 NM_009491.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026216.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026216.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026216.mm.1 NM_009491.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026211.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026211.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026211.mm.1 NM_009491.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026213.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026213.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026213.mm.1 NM_009491.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026212.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026212.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026212.mm.1 NM_009491.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026214.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026214.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026214.mm.1 NM_009491.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026217.mm.1 TC0500002960.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026217.mm.1 NM_009491.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026217.mm.1 NM_009491.3 Refseq
          Mouse Expression Set 430 1421701_PM_at - -
          Mouse Expression Set 430 1421701_at - -
          Murine Genome U74v2 Set 100679_at - -
          Murine Genome U74v2 Set 100679_PM_at - -
          Mouse Genome 430A 2.0 Array 1421701_PM_at - -
          Mouse Genome 430A 2.0 Array 1421701_at - -
          Affymetrix HT Mouse Genome 430A Array 1421701_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421701_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3190292 - -
          Illumina mouse-6 v1.0 expression beadchip 3190292 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2479412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2479412 - -
          mouse-6 v1.1 (Illumina) scl26242.5.1_33-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190292 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26242.5.1_33-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190292 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2479412 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2479412 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline