Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03807119_mH
          See other KLRA8 GE Assays ›
          Gene Symbol:
          Klra8
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 8
          Gene Aliases:
          Cmv-1, Cmv1, Ly49h, Ly49u<129>
          Chromosome Location:
          Chr.6: 130115226 - 130129898 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03807119_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101620.1 NP_001095090.1 4 - 5 565 89
          NM_010650.3 NP_034780.1 493 89
          GenBank mRNA AK143236.1 - 494 89
          L78253.1 - 411 89
          U12889.1 - 4 - 5 616 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003610.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra8

          Gene Ontology Categories:

          Function(s) Process(es)

          cell adhesion
          response to virus
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004527 NM_010650.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004527 AK143236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004527 NM_001101620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004527 U12889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121933 NM_010650.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121933 AK143236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121933 NM_001101620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P121933 U12889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004526 NM_010650.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004526 AK143236.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004526 NM_001101620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2004526 U12889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004526 NM_010650.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004526 AK143236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004526 NM_001101620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004526 U12889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121933 NM_010650.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121933 AK143236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121933 NM_001101620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P121933 U12889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004527 NM_010650.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004527 AK143236.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004527 NM_001101620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2004527 U12889.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98698 NM_010650.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98698 AK143236.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98698 L78253.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98698 NM_001101620.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P98698 U12889.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24846680 - -
          Whole Mouse Genome G4122A A_52_P98698 NM_010650.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98698 AK143236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98698 L78253.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98698 NM_001101620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98698 U12889.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127910 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114452 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105693 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121933 NM_010650.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121933 AK143236.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121933 NM_001101620.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121933 U12889.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24846675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548506 - -
          Affymetrix HT MG-430 PM Array Plate 1425417_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548504 - -
          Mouse Genome 430 2.0 Array 1425417_x_at - -
          Mouse Genome 430 2.0 Array 1425417_PM_x_at - -
          Affymetrix Murine Genome 6500A Array Msa13860_l_at - -
          Affymetrix Mouse Clariom S Array PSR0600025551.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025551.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025551.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025551.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025551.mm.1 NM_010650.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025554.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025554.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025554.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025554.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025554.mm.1 NM_010650.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025552.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025552.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025552.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025552.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025552.mm.1 NM_010650.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025555.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025555.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025555.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025555.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025555.mm.1 NM_010650.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025556.mm.1 TC0600003610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025556.mm.1 NM_001101620.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025556.mm.1 NM_001101620.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025556.mm.1 NM_010650.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025556.mm.1 NM_010650.3 Refseq
          Mouse Expression Set 430 1425436_x_at - -
          Mouse Expression Set 430 1425417_PM_x_at - -
          Mouse Expression Set 430 1425436_PM_x_at - -
          Mouse Expression Set 430 1425417_x_at - -
          Murine Genome U74v2 Set 93894_PM_f_at - -
          Murine Genome U74v2 Set 93894_f_at - -
          Murine Genome U74v2 Set 100326_f_at - -
          Murine Genome U74v2 Set 94779_f_at - -
          Murine Genome U74v2 Set 100326_PM_f_at - -
          Murine Genome U74v2 Set 94779_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1425436_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425436_x_at - -
          Mouse Genome 430A 2.0 Array 1425417_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425417_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425417_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425417_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610736 - -
          Illumina mouse-6 v1.0 expression beadchip 3610736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252006 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252006 - -
          mouse-6 v1.1 (Illumina) scl0016639.1_234-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0016639.1_234-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718242 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0016639.1_234-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline