Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03807525_s1
          See other SLC25A2 GE Assays ›
          Gene Symbol:
          Slc25a2
          Gene Name:
          solute carrier family 25 (mitochondrial carrier, ornithine transporter) member 2
          Gene Aliases:
          Ornt2
          Chromosome Location:
          Chr.18: 37637378 - 37638723 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03807525_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc25a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159275.1 NP_001152747.1 1 950 107
          GenBank mRNA AF332006.1 - 1 808 107
          AK077159.1 - 1 951 107
          AY583456.1 - 1 935 107
          BC114348.1 - 1 792 40051484 107
          BC125010.1 - 1 792 40051483 107

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102185 AY583456.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102185 BC114348.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102185 BC125010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102185 AK077159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102185 NM_001159275.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 75438 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158093 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102185 AY583456.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102185 BC114348.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102185 BC125010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102185 AK077159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2102185 NM_001159275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285279 AY583456.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285279 AK077159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285279 NM_001159275.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P285279 AF332006.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458422 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458420 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458421 - -
          Affymetrix HT MG-430 PM Array Plate 1425013_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458419 - -
          Mouse Genome 430 2.0 Array 1425013_PM_at - -
          Mouse Genome 430 2.0 Array 1425013_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83885_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83885_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800008890.mm.1 NM_001159275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008890.mm.1 NM_001159275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008890.mm.1 TC1800001206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008892.mm.1 NM_001159275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008892.mm.1 NM_001159275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008892.mm.1 TC1800001206.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008891.mm.1 NM_001159275.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008891.mm.1 NM_001159275.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008891.mm.1 TC1800001206.mm.1 MTA TC
          Mouse Expression Set 430 1425013_PM_at - -
          Mouse Expression Set 430 1425013_at - -
          Mouse Genome 430A 2.0 Array 1425013_PM_at - -
          Mouse Genome 430A 2.0 Array 1425013_at - -
          Affymetrix HT Mouse Genome 430A Array 1425013_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425013_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 394124 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 103290435 - -
          MouseWG-6 v2 Expression BeadChip 105890673 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2543036 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2653772 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0