Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04209268_g1
          See other 1700123K08RIK GE Assays ›
          Gene Symbol:
          1700123K08Rik
          Gene Name:
          RIKEN cDNA 1700123K08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 138559773 - 138565561 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04209268_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700123K08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029693.2 NP_083969.1 5 - 6 882 78
          GenBank mRNA AK007254.1 - 5 - 6 883 78
          BC147525.1 - 5 - 6 870 9056517 78
          BC147536.1 - 5 - 6 870 9056527 78

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700123K08Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 BC147525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 BC147536.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 NM_029693.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 AK007254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 BC147525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 BC147536.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 NM_029693.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P219594 AK007254.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139295 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 BC147525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 BC147536.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 NM_029693.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 AK007254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 BC147525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 BC147536.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 NM_029693.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P219594 AK007254.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24130708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24130729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24130706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24130737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24130696 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139295 - -
          Whole Mouse Genome G4122A A_51_P219594 BC147525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 BC147536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 NM_029693.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 AK007254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 BC147525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 BC147536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 NM_029693.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P219594 AK007254.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24130681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04895375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04896020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04448933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04297401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04448957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04448961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04448948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24130668 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10535032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10535033 - -
          Affymetrix HT MG-430 PM Array Plate 1421675_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10535027 - -
          Mouse Genome 430 2.0 Array 1421675_at - -
          Mouse Genome 430 2.0 Array 1421675_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76658_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76658_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500031803.mm.1 TC0500003480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031803.mm.1 NM_029693.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031803.mm.1 NM_029693.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031805.mm.1 TC0500003480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031805.mm.1 NM_029693.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031805.mm.1 NM_029693.2 Refseq
          Mouse Expression Set 430 1421675_PM_at - -
          Mouse Expression Set 430 1421675_at - -
          Mouse Genome 430A 2.0 Array 1421675_PM_at - -
          Mouse Genome 430A 2.0 Array 1421675_at - -
          Affymetrix HT Mouse Genome 430A Array 1421675_at - -
          Affymetrix HT Mouse Genome 430A Array 1421675_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 674576 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2850025 - -
          MouseWG-6 v2 Expression BeadChip 360373 - -
          Illumina mouse-6 v1.0 expression beadchip 103610239 - -
          Illumina mouse-6 v1.0 expression beadchip 360373 - -
          Illumina mouse-6 v1.0 expression beadchip 2850025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2760476 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2648526 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2760476 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259327 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2648526 - -
          mouse-6 v1.1 (Illumina) GI_13386431-S - -
          mouse-6 v1.1 (Illumina) scl076658.2_162-S - -
          mouse-6 v1.1 (Illumina) scl0076658.1_124-S - -
          Illumina Sentrix mouse-6.v1 bead array 103610239 - -
          Illumina Sentrix mouse-6.v1 bead array 360373 - -
          Illumina Sentrix mouse-6.v1 bead array 2850025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0076658.1_124-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl076658.2_162-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 103610239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2760476 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2648526 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2760476 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2648526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline