Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04210574_g1
          See other OLFR1115 GE Assays ›
          Gene Symbol:
          Olfr1115
          Gene Name:
          olfactory receptor 1115
          Gene Aliases:
          MOR264-20, MOR273-4P, Olfr1530-ps1
          Chromosome Location:
          Chr.2: 87251840 - 87252941 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04210574_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146297.2 NP_666409.2 1 1038 101
          GenBank mRNA BC141024.1 - 1 1038 9055636 101

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1115

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992734 BC141024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992734 NM_146297.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992735 BC141024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992735 NM_146297.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992735 BC141024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992735 NM_146297.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992734 BC141024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992734 NM_146297.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145340 BC141024.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145340 NM_146297.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21405319 - -
          Whole Mouse Genome G4122A A_51_P145340 BC141024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145340 NM_146297.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01361006 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01447298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01360998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21405331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01361007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01447297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01207034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01835732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01174311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01207039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21405289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473520 - -
          Affymetrix Mouse Gene 1.1 ST Array 10473519 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 390618 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2900278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2777179 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615222 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615222 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2777179 - -
          mouse-6 v1.1 (Illumina) scl0258294.1_254-S - -
          Illumina Sentrix mouse-6.v1 bead array 2900278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258294.1_254-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900278 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777179 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline