Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04211970_m1
          See other GPATCH3 GE Assays ›
          Gene Symbol:
          Gpatch3
          Gene Name:
          G patch domain containing 3
          Gene Aliases:
          D930035B09Rik, Gpatc3
          Chromosome Location:
          Chr.4: 133574745 - 133584242 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04211970_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpatch3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172876.2 NP_766464.1 4 - 5 1130 93
          GenBank mRNA AK053051.1 - 4 - 5 1131 93
          BC096468.1 - 4 - 5 1113 3665952 93
          BC121828.1 - 4 - 5 1128 40107316 93
          BC121829.1 - 4 - 5 1105 40107315 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001614.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpatch3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          nucleic acid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318320 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259555 AK053051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259555 NM_172876.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259555 AK053051.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259555 NM_172876.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204831 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55922 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134583 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259555 AK053051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259555 NM_172876.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259555 AK053051.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259555 NM_172876.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23318298 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134583 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05409728 - -
          Whole Mouse Genome G4122A A_51_P259555 AK053051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259555 NM_172876.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259555 AK053051.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259555 NM_172876.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P188309 AK050179.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126404 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134583 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03392081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03392077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03865135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03865139 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03865146 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03865154 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03865137 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318320 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23318265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23318298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03865154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03865150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03935918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04096720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03392082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03202524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03392077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03865137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04023527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03259701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04096718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03457177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03392081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04004555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03422191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03865146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23318290 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10508896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508899 - -
          Affymetrix HT MG-430 PM Array Plate 1440406_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10508892 - -
          Mouse Genome 430 2.0 Array 1440406_PM_at - -
          Mouse Genome 430 2.0 Array 1440406_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242691_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 242691_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400012795.mm.1 NM_172876.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012795.mm.1 NM_172876.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012795.mm.1 TC0400001614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012792.mm.1 NM_172876.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012792.mm.1 NM_172876.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012792.mm.1 TC0400001614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012793.mm.1 NM_172876.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012793.mm.1 NM_172876.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012793.mm.1 TC0400001614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012796.mm.1 NM_172876.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012796.mm.1 NM_172876.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012796.mm.1 TC0400001614.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400012794.mm.1 NM_172876.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012794.mm.1 NM_172876.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400012794.mm.1 TC0400001614.mm.1 MTA TC
          Mouse Expression Set 430 1440406_at - -
          Mouse Expression Set 430 1440406_PM_at - -
          Murine Genome U74v2 Set 138459_PM_at - -
          Murine Genome U74v2 Set 138459_at - -
          Affymetrix HT Mouse Genome 430B Array 1440406_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440406_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 395475 - -
          ABI Mouse Genome Survey Microarray 637938 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7040110 - -
          Illumina mouse-6 v1.0 expression beadchip 101770538 - -
          Illumina mouse-6 v1.0 expression beadchip 7040110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694857 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694857 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249324 - -
          mouse-6 v1.1 (Illumina) scl24859.6.20_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040110 - -
          Illumina Sentrix mouse-6.v1 bead array 101770538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24859.6.20_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040110 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101770538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2694857 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2962312 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2562780 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2962312 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694857 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline