Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm04214657_uH
          See other KLRA16 GE Assays ›
          Gene Symbol:
          Klra16
          Gene Name:
          killer cell lectin-like receptor, subfamily A, member 16
          Gene Aliases:
          Klra24, Ly-49P, Ly49P, Ly49x
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm04214657_uH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klra16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013794.4 NP_038822.3 7 1050 127
          GenBank mRNA AF146570.4 - 7 1050 127

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klra16

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958039 BC144940.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958039 AF146570.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958039 BC119242.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958039 BC144924.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958039 NM_013794.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958038 AF146570.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1958038 NM_013794.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2028734 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2028734 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958038 AF146570.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958038 NM_013794.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958039 BC144940.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958039 AF146570.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958039 BC119242.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958039 BC144924.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1958039 NM_013794.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662054 AF146570.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662054 NM_013794.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662054 AF074458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662054 AF074461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P662054 AF074462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434836 BC144940.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434836 AF218082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434836 AF146570.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434836 AF218080.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434836 AF218081.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10548486 - -
          Affymetrix HT MG-430 PM Array Plate 1425959_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1420790_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10548455 - -
          Mouse Genome 430 2.0 Array 1425959_x_at - -
          Mouse Genome 430 2.0 Array 1420790_x_at - -
          Mouse Genome 430 2.0 Array 1420790_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425959_PM_x_at - -
          Mouse Expression Set 430 1425959_PM_x_at - -
          Mouse Expression Set 430 1420790_PM_x_at - -
          Mouse Expression Set 430 1420790_x_at - -
          Mouse Expression Set 430 1425959_x_at - -
          Murine Genome U74v2 Set 95908_PM_at - -
          Murine Genome U74v2 Set 95908_at - -
          Mouse Genome 430A 2.0 Array 1425959_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1420790_x_at - -
          Mouse Genome 430A 2.0 Array 1420790_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425959_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420790_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420790_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425959_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425959_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 392370 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106400541 - -
          Illumina mouse-6 v1.0 expression beadchip 106400541 - -
          Illumina mouse-6 v1.0 expression beadchip 3360167 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360167 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222914 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229795 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222914 - -
          mouse-6 v1.1 (Illumina) GI_9256548-S - -
          mouse-6 v1.1 (Illumina) scl0027424.1_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 106400541 - -
          Illumina Sentrix mouse-6.v1 bead array 3360167 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0027424.1_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360167 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106400541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2724548 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3005175 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229795 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3005175 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2724548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline