Search
Search

Itln1| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010584.3 | NP_034714.1 | 5 - 6 | 637 | 77 | |
| GenBank mRNA | AK007447.1 | - | 5 - 6 | 637 | 77 | |
| AY157364.1 | - | 5 - 6 | 621 | 77 | ||
| AY619693.1 | - | 5 - 6 | 620 | 77 | ||
Set Membership: |
|