Search
Search
| Gene Symbol: Tnni2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017185.1 | NP_058881.1 | 4 - 5 | 220 | 62 | |
| GenBank mRNA | FQ216337.1 | - | 4 - 5 | 262 | 62 | |
| FQ217026.1 | - | 5 - 6 | 262 | 62 | ||
| FQ217454.1 | - | 5 - 6 | 270 | 62 | ||
| FQ217985.1 | - | 5 - 6 | 262 | 62 | ||
| FQ218058.1 | - | 4 - 5 | 294 | 62 | ||
| FQ223581.1 | - | 5 - 6 | 262 | 62 | ||
| FQ223772.1 | - | 5 - 6 | 268 | 62 | ||
| FQ224443.1 | - | 5 - 6 | 263 | 62 | ||
| FQ224464.1 | - | 5 - 6 | 262 | 62 | ||
| FQ224489.1 | - | 5 - 6 | 262 | 62 | ||
| M73701.1 | - | 4 - 5 | 220 | 62 | ||
Set Membership: |
|