Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00452692_m1
          See other PPIL4 GE Assays ›
          Gene Symbol:
          Ppil4
          Gene Name:
          peptidylprolyl isomerase (cyclophilin)-like 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1897316 - 1931181 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00452692_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppil4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108457.1 NP_001101927.1 10 - 11 1012 93
          XM_006227621.2 XP_006227683.1 10 - 11 1046 93
          GenBank mRNA FQ225576.1 - 10 - 11 1051 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000025.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          protein folding
          regulation of phosphorylation of RNA polymerase II C-terminal domain
          nucleotide binding
          peptidyl-prolyl cis-trans isomerase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P653448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 708379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515504 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545169 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172715 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83851 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P653448 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P653448 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P653448 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P538725 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66443 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10701736 - -
          Affymetrix Rat Gene 1.0 ST Array 10701745 - -
          Affymetrix Rat Gene 1.0 ST Array 10701738 - -
          Affymetrix Rat Gene 1.0 ST Array 10701741 - -
          Affymetrix Rat Gene 1.0 ST Array 10701740 - -
          Affymetrix Rat Gene 1.0 ST Array 10701744 - -
          Affymetrix Rat Gene 1.0 ST Array 10701747 - -
          Affymetrix Rat Gene 1.0 ST Array 10701737 - -
          Affymetrix Rat Gene 1.0 ST Array 10701743 - -
          Affymetrix Rat Gene 1.0 ST Array 10701746 - -
          Affymetrix Rat Gene 1.0 ST Array 10701739 - -
          Affymetrix Rat Gene 1.0 ST Array 10701735 - -
          Affymetrix Rat Gene 1.0 ST Array 10701742 - -
          Affymetrix Rat Gene 1.1 ST Array 10701734 - -
          Rat Genome 230 2.0 Array 1375061_PM_at - -
          Rat Genome 230 2.0 Array 1396171_at - -
          Rat Genome 230 2.0 Array 1396171_PM_at - -
          Rat Genome 230 2.0 Array 1375061_at - -
          Affymetrix HT Rat Focus Array 1375061_at - -
          Affymetrix HT Rat Focus Array 1375061_PM_at - -
          Affymetrix HT Rat Focus Array 1396171_at - -
          Affymetrix HT Rat Focus Array 1396171_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100000165.rn.1 TC0100000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000166.rn.1 TC0100000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000163.rn.1 TC0100000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000167.rn.1 TC0100000025.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100000168.rn.1 TC0100000025.rn.1 RTA TC
          Rat Expression Set 230 1375061_at - -
          Rat Expression Set 230 1375061_PM_at - -
          Rat Expression Set 230 1396171_PM_at - -
          Rat Expression Set 230 1396171_at - -
          Affymetrix Rat Gene 1.0 ST Array 10701734 - -
          Rat Genome U34 Set rc_AI069975_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21775823 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline