Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00479916_m1
          See other HSD17B12 GE Assays ›
          Gene Symbol:
          Hsd17b12
          Gene Name:
          hydroxysteroid (17-beta) dehydrogenase 12
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 82924699 - 83048456 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00479916_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsd17b12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_032066.1 NP_114455.1 6 - 7 514 116
          XM_006234599.2 XP_006234661.1 6 - 7 681 116
          GenBank mRNA BC061543.1 - 6 - 7 535 6888949 116
          FQ213557.1 - 6 - 7 550 116
          FQ214777.1 - 6 - 7 579 116
          U81186.1 - 6 - 7 514 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003713.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsd17b12

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid biosynthetic process
          estrogen biosynthetic process
          biological_process
          positive regulation of cell-substrate adhesion
          extracellular matrix organization
          oxidation-reduction process
          fibronectin binding
          molecular_function
          estradiol 17-beta-dehydrogenase activity
          collagen binding
          heparin binding
          oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P517896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 110776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 67755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 634848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 290422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 448898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741000 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947472 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191075 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109659 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 171991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127943 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168568 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157247 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 129253 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156756 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115047 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517896 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517896 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P517896 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517896 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700309975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4023 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97519 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5857 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 425 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48113 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92196 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53959 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48073 - -
          Agilent Rat cDNA Microarray (G4105A) 700309975 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847655 - -
          Affymetrix Rat Gene 1.0 ST Array 10847658 - -
          Affymetrix Rat Gene 1.0 ST Array 10847654 - -
          Affymetrix Rat Gene 1.0 ST Array 10847649 - -
          Affymetrix Rat Gene 1.0 ST Array 10847653 - -
          Affymetrix Rat Gene 1.0 ST Array 10847648 - -
          Affymetrix Rat Gene 1.0 ST Array 10847656 - -
          Affymetrix Rat Gene 1.0 ST Array 10847652 - -
          Affymetrix Rat Gene 1.0 ST Array 10847647 - -
          Affymetrix Rat Gene 1.0 ST Array 10847650 - -
          Affymetrix Rat Gene 1.0 ST Array 10847651 - -
          Affymetrix Rat Gene 1.0 ST Array 10847657 - -
          Affymetrix Rat Gene 1.1 ST Array 10847646 - -
          Rat Genome 230 2.0 Array 1368051_PM_at - -
          Rat Genome 230 2.0 Array 1368051_at - -
          Affymetrix HT Rat Focus Array 1368051_at - -
          Affymetrix HT Rat Focus Array 1368051_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300028328.rn.1 TC0300003713.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300028329.rn.1 TC0300003713.rn.1 RTA TC
          Rat Expression Set 230 1368051_at - -
          Rat Expression Set 230 1368051_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847646 - -
          Rat Genome U34 Set U81186_g_at - -
          Rat Genome U34 Set U81186_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22056549 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372600 - -
          RatRef-12 Expression BeadChip ILMN_51036 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline