Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00515970_m1
          See other HEPH GE Assays ›
          Gene Symbol:
          Heph
          Gene Name:
          hephaestin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 65377056 - 65658479 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00515970_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Heph
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133304.1 NP_579838.1 7 - 8 1315 75
          XM_006257046.2 XP_006257108.1 8 - 9 1615 75
          XM_006257048.2 XP_006257110.1 8 - 9 1850 75
          XM_006257049.2 XP_006257111.1 7 - 8 1650 75
          XM_008773248.1 XP_008771470.1 9 - 10 2248 75
          XM_008773249.1 XP_008771471.1 9 - 10 1944 75
          XM_008773250.1 XP_008771472.1 8 - 9 1618 75
          GenBank mRNA AF246120.1 - 7 - 8 1315 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000554.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Heph

          Panther Classification:

          Molecular Function -

          oxidase

          Gene Ontology Categories:

          Function(s) Process(es)

          copper ion transport
          iron ion transport
          cellular iron ion homeostasis
          erythrocyte differentiation
          oxidation-reduction process
          ferroxidase activity
          copper ion binding
          ferrous iron binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1051724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 419900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664889 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908394 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 599912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458158 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 722769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679107 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190021 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92850 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72229 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150350 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167969 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146364 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 156477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1051724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1051724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1051724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1051724 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 22460 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10048 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25649 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14473 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101920 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41748 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94322 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934081 - -
          Affymetrix Rat Gene 1.0 ST Array 10934093 - -
          Affymetrix Rat Gene 1.0 ST Array 10934077 - -
          Affymetrix Rat Gene 1.0 ST Array 10934075 - -
          Affymetrix Rat Gene 1.0 ST Array 10934090 - -
          Affymetrix Rat Gene 1.0 ST Array 10934080 - -
          Affymetrix Rat Gene 1.0 ST Array 10934092 - -
          Affymetrix Rat Gene 1.0 ST Array 10934079 - -
          Affymetrix Rat Gene 1.0 ST Array 10934082 - -
          Affymetrix Rat Gene 1.0 ST Array 10934094 - -
          Affymetrix Rat Gene 1.0 ST Array 10934089 - -
          Affymetrix Rat Gene 1.0 ST Array 10934074 - -
          Affymetrix Rat Gene 1.0 ST Array 10934078 - -
          Affymetrix Rat Gene 1.0 ST Array 10934086 - -
          Affymetrix Rat Gene 1.0 ST Array 10934076 - -
          Affymetrix Rat Gene 1.0 ST Array 10934087 - -
          Affymetrix Rat Gene 1.0 ST Array 10934091 - -
          Affymetrix Rat Gene 1.0 ST Array 10934085 - -
          Affymetrix Rat Gene 1.0 ST Array 10934083 - -
          Affymetrix Rat Gene 1.0 ST Array 10934084 - -
          Affymetrix Rat Gene 1.1 ST Array 10934073 - -
          Rat Genome 230 2.0 Array 1368533_at - -
          Rat Genome 230 2.0 Array 1368533_PM_at - -
          Affymetrix HT Rat Focus Array 1368533_PM_at - -
          Affymetrix HT Rat Focus Array 1368533_at - -
          Affymetrix Rat Clariom S Assay PSR0X00003458.rn.1 TC0X00000554.rn.1 RTA TC
          Rat Expression Set 230 1368533_PM_at - -
          Rat Expression Set 230 1368533_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934073 - -
          Rat Genome U34 Set rc_AI031036_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21129618 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371544 - -
          RatRef-12 Expression BeadChip ILMN_51870 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline