Search
Search
| Gene Symbol: Mylpf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012605.2 | NP_036737.1 | 5 - 6 | 403 | 57 | |
| GenBank mRNA | BI278596.1 | - | 2 - 3 | 57 | ||
| FQ215085.1 | - | 5 - 6 | 436 | 57 | ||
| FQ216934.1 | - | 5 - 6 | 419 | 57 | ||
| FQ217090.1 | - | 5 - 6 | 421 | 57 | ||
| FQ217098.1 | - | 5 - 6 | 419 | 57 | ||
| FQ217115.1 | - | 3 - 4 | 268 | 57 | ||
| FQ217155.1 | - | 5 - 6 | 419 | 57 | ||
| FQ217219.1 | - | 5 - 6 | 419 | 57 | ||
| FQ217259.1 | - | 5 - 6 | 419 | 57 | ||
| FQ217349.1 | - | 4 - 5 | 343 | 57 | ||
| FQ217365.1 | - | 5 - 6 | 418 | 57 | ||
| FQ217368.1 | - | 5 - 6 | 435 | 57 | ||
| FQ217533.1 | - | 5 - 6 | 447 | 57 | ||
| FQ217703.1 | - | 5 - 6 | 419 | 57 | ||
| FQ217740.1 | - | 5 - 6 | 435 | 57 | ||
| FQ217881.1 | - | 5 - 6 | 416 | 57 | ||
| FQ217944.1 | - | 5 - 6 | 419 | 57 | ||
| FQ218017.1 | - | 5 - 6 | 419 | 57 | ||
| FQ223762.1 | - | 5 - 6 | 400 | 57 | ||
| FQ223868.1 | - | 5 - 6 | 419 | 57 | ||
| FQ223911.1 | - | 5 - 6 | 435 | 57 | ||
| FQ223944.1 | - | 4 - 5 | 285 | 57 | ||
| FQ223946.1 | - | 5 - 6 | 419 | 57 | ||
| FQ223966.1 | - | 5 - 6 | 419 | 57 | ||
| FQ223985.1 | - | 5 - 6 | 419 | 57 | ||
| FQ224150.1 | - | 3 - 4 | 262 | 57 | ||
| FQ224231.1 | - | 5 - 6 | 419 | 57 | ||
| FQ224253.1 | - | 5 - 6 | 418 | 57 | ||
| FQ224391.1 | - | 5 - 6 | 417 | 57 | ||
| FQ224404.1 | - | 5 - 6 | 417 | 57 | ||
| FQ224511.1 | - | 5 - 6 | 419 | 57 | ||
| FQ224525.1 | - | 5 - 6 | 438 | 57 | ||
| FQ224720.1 | - | 5 - 6 | 419 | 57 | ||
| FQ224781.1 | - | 5 - 6 | 435 | 57 | ||
| J00754.1 | - | 2 - 3 | 155 | 57 | ||
Set Membership: |
|